Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JUNB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002229.2 | NP_002220.1 | 1 | 203 | 105 | |
GenBank mRNA | AK222532.1 | - | 1 | 222 | 105 | |
AK313737.1 | - | 1 | 214 | 105 | ||
BC004250.1 | - | 1 | 203 | 3621911 | 105 | |
BC009465.1 | - | 1 | 203 | 3611163 | 105 | |
BC009466.1 | - | 1 | 203 | 3611749 | 105 | |
BC110372.1 | - | 1 | 205 | 105 | ||
DQ650707.1 | - | 1 | 105 | |||
X51345.1 | - | 1 | 180 | 105 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |