Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05002045_s1
          See other LOC100130920 GE Assays ›
          Gene Symbol:
          LOC100130920 PRKCQ
          Gene Name:
          uncharacterized LOC100130920
          protein kinase C theta
          Gene Aliases:
          -
          PRKCT, nPKC-theta
          Chromosome Location:
          -
          Chromosome Location:
          Chr.10: 6393038 - 6580646 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs05002045_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100130920
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK093639.1 - 1 1724 133
          Gene Symbol: PRKCQ
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006717465.3 XP_006717528.1 1 1724 133
          XM_017016410.1 XP_016871899.1 1 1724 133
          XM_017016411.1 XP_016871900.1 1 1724 133

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_006717465.3, XM_017016410.1, XM_017016411.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          response to hypoxia
          regulation of transcription, DNA-templated
          membrane protein ectodomain proteolysis
          cellular component disassembly involved in execution phase of apoptosis
          inflammatory response
          axon guidance
          aging
          response to heat
          response to glucose
          regulation of G2/M transition of mitotic cell cycle
          response to organic cyclic compound
          peptidyl-serine phosphorylation
          regulation of vasoconstriction
          platelet activation
          positive regulation of telomere maintenance via telomerase
          positive regulation of interleukin-17 production
          positive regulation of interleukin-4 production
          response to insulin
          intracellular signal transduction
          Fc-epsilon receptor signaling pathway
          positive regulation of T cell proliferation
          tissue regeneration
          respiratory burst
          negative regulation of insulin receptor signaling pathway
          positive regulation of protein secretion
          T cell receptor signaling pathway
          positive regulation of T cell activation
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of filopodium assembly
          positive regulation of stress fiber assembly
          positive regulation of telomerase activity
          cell chemotaxis
          negative regulation of T cell apoptotic process
          regulation of platelet aggregation
          positive regulation of NIK/NF-kappaB signaling
          positive regulation of telomere capping
          positive regulation of T-helper 17 type immune response
          positive regulation of T-helper 2 cell activation
          protein kinase activity
          protein serine/threonine kinase activity
          protein kinase C activity
          protein binding
          ATP binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0X00009385.hg.1 ENST00000408539.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009385.hg.1 ENST00000408539.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009385.hg.1 NR_039674.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009385.hg.1 NR_039674.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009385.hg.1 ENST00000408539.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009385.hg.1 ENST00000408539.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009385.hg.1 NR_039674.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009385.hg.1 NR_039674.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0