Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000484438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000484438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000484438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000484438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000484438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000484438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |