Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04963435_s1
          See other TAB1 GE Assays ›
          Gene Symbol:
          TAB1
          Gene Name:
          TGF-beta activated kinase 1/MAP3K7 binding protein 1
          Gene Aliases:
          3'-Tab1, MAP3K7IP1
          Chromosome Location:
          Chr.22: 39399754 - 39437127 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04963435_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TAB1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006116.2 NP_006107.1 11 2826 70
          GenBank mRNA BC038582.1 - 11 2801 70
          BC050554.1 - 11 2839 6202841 70
          EF036484.1 - 1 122 70
          U49928.1 - 11 2804 70

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TAB1

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          stimulatory C-type lectin receptor signaling pathway
          MyD88-dependent toll-like receptor signaling pathway
          heart morphogenesis
          cardiac septum development
          protein dephosphorylation
          transforming growth factor beta receptor signaling pathway
          I-kappaB kinase/NF-kappaB signaling
          JNK cascade
          lung development
          aorta development
          Fc-epsilon receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          coronary vasculature development
          nucleotide-binding oligomerization domain containing signaling pathway
          protein serine/threonine phosphatase activity
          protein binding
          enzyme activator activity
          kinase activator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000461775.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000461775.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000461775.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000461775.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000473491.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000473491.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000473491.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000473491.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000473613.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000473613.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000473613.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000473613.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000473613.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000488859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000488859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000488859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000488859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000490819.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000490819.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000490819.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 ENST00000490819.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NR_040535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NR_040535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NR_040535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NR_040535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NR_040535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NR_040535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NR_040535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007401.hg.1 NR_040535.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000216160.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000331454.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000461775.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000461775.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000461775.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000461775.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000473491.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000473491.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000473491.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000473491.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000473613.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000473613.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000473613.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000473613.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000473613.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000488859.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000488859.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000488859.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000488859.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000490819.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000490819.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000490819.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 ENST00000490819.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_006116.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NM_153497.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NR_040535.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NR_040535.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NR_040535.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NR_040535.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NR_040535.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NR_040535.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NR_040535.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007401.hg.1 NR_040535.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0