Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007629.hg.1 |
ENST00000563187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007629.hg.1 |
ENST00000563187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007629.hg.1 |
ENST00000567922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007629.hg.1 |
NR_130771.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007629.hg.1 |
NR_130771.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007629.hg.1 |
NR_130771.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007629.hg.1 |
NR_130771.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007629.hg.1 |
NR_130771.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007629.hg.1 |
NR_130772.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007629.hg.1 |
NR_130772.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007629.hg.1 |
NR_130772.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007629.hg.1 |
NR_130772.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015569.hg.1 |
ENST00000615763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015569.hg.1 |
ENST00000615763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015569.hg.1 |
ENST00000615763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015569.hg.1 |
ENST00000617368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015569.hg.1 |
ENST00000617368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015569.hg.1 |
ENST00000617368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015569.hg.1 |
ENST00000617368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015569.hg.1 |
ENST00000617368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015569.hg.1 |
ENST00000615763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015569.hg.1 |
ENST00000615763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015569.hg.1 |
ENST00000615763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015569.hg.1 |
ENST00000617368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015569.hg.1 |
ENST00000617368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015569.hg.1 |
ENST00000617368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015569.hg.1 |
ENST00000617368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015569.hg.1 |
ENST00000617368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007629.hg.1 |
ENST00000563187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007629.hg.1 |
ENST00000563187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007629.hg.1 |
ENST00000567922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007629.hg.1 |
NR_130771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007629.hg.1 |
NR_130771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007629.hg.1 |
NR_130771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007629.hg.1 |
NR_130771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007629.hg.1 |
NR_130771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007629.hg.1 |
NR_130772.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007629.hg.1 |
NR_130772.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007629.hg.1 |
NR_130772.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007629.hg.1 |
NR_130772.1 |
Refseq |