Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04941623_m1
          See other CRYBB2P1 GE Assays ›
          Gene Symbol:
          CRYBB2P1
          Gene Name:
          crystallin beta B2 pseudogene 1
          Gene Aliases:
          CRYB2B
          Chromosome Location:
          Chr.22: 25448087 - 25461678 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04941623_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CRYBB2P1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033733.1 1 - 2 203 114
          GenBank mRNA BC047380.1 - 1 - 2 206 114
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DW426763.1 - 1 - 2 202 Chr. 22 - 25448105 - 25453399 [+] on Build GRCh38 114
          GenBankmRNA T09052.1 - 169 Chr. - - [-] on Build 113
          GenBankmRNA DA491429.1 - 1 - 2 185 Chr. 22 - 25448105 - 25459439 [+] on Build GRCh38 114
          GenBankmRNA DA593787.1 - 1 - 2 204 Chr. 22 - 25448087 - 25457376 [+] on Build GRCh38 115
          GenBankmRNA BU609031.1 - 3 - 4 Chr. 22 - 25448154 - 25459677 [-] on Build GRCh38 114
          GenBankmRNA DA778799.1 - 1 - 2 190 Chr. 22 - 25448100 - 25457394 [+] on Build GRCh38 114
          GenBankmRNA BG432219.1 - 1 - 2 195 Chr. 22 - 25448096 - 25457398 [+] on Build GRCh38 114
          GenBankmRNA BX505859.1 - 1 - 2 195 Chr. 22 - 25448096 - 25457350 [+] on Build GRCh38 114
          GenBankmRNA BX325511.2 - 1 - 2 80 Chr. 22 - 25448214 - 25461054 [+] on Build GRCh38 115
          GenBankmRNA CR746663.1 - 1 - 2 184 Chr. 22 - 25448117 - 25457330 [+] on Build GRCh38 114
          GenBankmRNA BI225025.1 - 1 - 2 175 Chr. 22 - 25448115 - 25459471 [+] on Build GRCh38 114
          GenBankmRNA BQ433813.1 - 1 - 2 172 Chr. 22 - 25448118 - 25459706 [+] on Build GRCh38 114
          GenBankmRNA BX395320.2 - 4 - 5 547 Chr. 22 - 25436698 - 25455822 [+] on Build GRCh38 113
          GenBankmRNA BE796362.1 - 1 - 2 171 Chr. 22 - 25448119 - 25459661 [+] on Build GRCh38 114
          GenBankmRNA CX751973.1 - 1 - 2 170 Chr. 22 - 25448121 - 25460946 [+] on Build GRCh38 114
          GenBankmRNA BU959180.1 - 1 - 2 188 Chr. 22 - 25448105 - 25459594 [+] on Build GRCh38 114
          GenBankmRNA BI667140.1 - 1 - 2 211 Chr. 22 - 25448080 - 25457330 [+] on Build GRCh38 114
          GenBankmRNA AL110341.1 - 1 - 2 190 Chr. 22 - 25448100 - 25453400 [+] on Build GRCh38 114
          GenBankmRNA BM789420.1 - 1 - 2 161 Chr. 22 - 25448129 - 25459442 [+] on Build GRCh38 114
          GenBankmRNA BM928787.1 - 175 Chr. - - [-] on Build 114
          GenBankmRNA DB162321.1 - 1 - 2 150 Chr. 22 - 25448140 - 25459476 [+] on Build GRCh38 114
          GenBankmRNA DA104688.1 - 1 - 2 185 Chr. 22 - 25448105 - 25457374 [+] on Build GRCh38 114
          GenBankmRNA AW963545.1 - 1 - 2 129 Chr. 22 - 25448161 - 25459520 [+] on Build GRCh38 114
          GenBankmRNA BX388339.2 - 2 - 3 Chr. 22 - 25436704 - 25455735 [-] on Build GRCh38 114
          GenBankmRNA BX952640.1 - 1 - 2 186 Chr. 22 - 25448105 - 25459505 [+] on Build GRCh38 114
          GenBankmRNA BI758640.1 - 1 - 2 210 Chr. 22 - 25448117 - 25460889 [+] on Build GRCh38 115
          GenBankmRNA DA571936.1 - 1 - 2 201 Chr. 22 - 25448089 - 25466100 [+] on Build GRCh38 114
          GenBankmRNA BQ651116.1 - 139 Chr. - - [-] on Build 114
          GenBankmRNA BM458453.1 - 1 - 2 173 Chr. 22 - 25448117 - 25459681 [+] on Build GRCh38 114
          GenBankmRNA DA494750.1 - 1 - 2 185 Chr. 22 - 25448105 - 25457407 [+] on Build GRCh38 114
          GenBankmRNA BI598484.1 - 1 - 2 211 Chr. 22 - 25448080 - 25459505 [+] on Build GRCh38 114
          GenBankmRNA BQ650704.1 - 1 - 2 149 Chr. 22 - 25448154 - 25461131 [+] on Build GRCh38 114
          GenBankmRNA BX387326.2 - 4 - 5 547 Chr. 22 - 25436698 - 25455822 [+] on Build GRCh38 113
          GenBankmRNA DB485488.1 - 1 - 2 150 Chr. 22 - 25448140 - 25457282 [+] on Build GRCh38 114

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream LDL receptor related protein 5 like LRP5L -42.7kb Anti-sense Chr. 22 - 25351418 - 25405410 on NCBI Build GRCh38
          Downstream microRNA 6817 MIR6817 hsa-mir-6817 2.2kb Sense Chr. 22 - 25455646 - 25455711 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000382734.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000382734.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000382734.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000382734.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000382734.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000415709.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000415709.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000415709.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000415709.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000415709.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000609084.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000609084.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 ENST00000609084.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 NR_033733.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 NR_033733.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 NR_033733.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 NR_033733.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 NR_033733.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 NR_033733.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 NR_033734.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 NR_033734.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 NR_033734.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 NR_033734.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200009237.hg.1 NR_033734.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000354451.6 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000382734.9 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000382734.9 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000382734.9 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000382734.9 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000382734.9 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000415709.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000415709.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000415709.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000415709.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000415709.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000609084.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000609084.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 ENST00000609084.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 NR_033733.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 NR_033733.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 NR_033733.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 NR_033733.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 NR_033733.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 NR_033733.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 NR_033734.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 NR_033734.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 NR_033734.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 NR_033734.1 Refseq
          Affymetrix Human Clariom D Assay TC2200009237.hg.1 NR_033734.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0