Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009002.hg.1 |
ENST00000526703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009002.hg.1 |
ENST00000526703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009002.hg.1 |
ENST00000526703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009002.hg.1 |
ENST00000526605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009002.hg.1 |
ENST00000526703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009002.hg.1 |
ENST00000526703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009002.hg.1 |
ENST00000526703.1 |
Ensembl |