Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000504820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000504820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000504820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000504820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000510128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000510128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011807.hg.1 |
ENST00000510128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000504820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000504820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000504820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000504820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000506486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000510128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000510128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011807.hg.1 |
ENST00000510128.1 |
Ensembl |