Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04409246_s1
          See other LOC101928944 GE Assays ›
          Gene Symbol:
          LOC101928944
          Gene Name:
          uncharacterized LOC101928944
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 80751200 - 80762803 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04409246_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC101928944
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_120570.1 1 523 112
          GenBank mRNA AY726583.1 - 1 523 112
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BI520577.1 - 501 Chr. - - [-] on Build 113

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream teneurin transmembrane protein 4 TENM4 Doc4,
          ETM5,
          ODZ4,
          TNM4,
          Ten-M4
          -1.3mb Sense Chr. 11 - 78653283 - 79441019 on NCBI Build GRCh38
          Downstream MIR4300 host gene MIR4300HG 1.1mb Sense Chr. 11 - 81879851 - 82403913 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100011771.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100011771.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011771.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011771.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011771.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011771.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011771.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011771.hg.1 NR_120570.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100011771.hg.1 NR_120570.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100011771.hg.1 NR_120570.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100011771.hg.1 NR_120570.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100011771.hg.1 NR_120570.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100011771.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100190964.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100190964.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100190964.hg.1 TC1100011771.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100190964.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100190964.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100190957.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100190957.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100190957.hg.1 TC1100011771.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100190957.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100190957.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100190961.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100190961.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100190961.hg.1 TC1100011771.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100190961.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100190961.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100190962.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100190962.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100190962.hg.1 TC1100011771.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100190962.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100190962.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay TC1100011771.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011771.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011771.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011771.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011771.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011771.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011771.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay TC1100011771.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay TC1100011771.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay TC1100011771.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay TC1100011771.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay TC1100011771.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100190963.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100190963.hg.1 NR_120570.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100190963.hg.1 TC1100011771.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100190963.hg.1 ENST00000526840.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100190963.hg.1 NR_120570.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0