Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012488.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012488.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012488.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012488.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012488.hg.1 |
NR_104182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012488.hg.1 |
NR_104182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012488.hg.1 |
NR_104182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012488.hg.1 |
NR_104182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191119.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191119.hg.1 |
NR_104182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191119.hg.1 |
TC0200012488.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200191119.hg.1 |
NR_104182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191119.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191121.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191121.hg.1 |
NR_104182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191121.hg.1 |
TC0200012488.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200191121.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191121.hg.1 |
NR_104182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191120.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191120.hg.1 |
NR_104182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191120.hg.1 |
TC0200012488.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200191120.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191120.hg.1 |
NR_104182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191122.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191122.hg.1 |
NR_104182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191122.hg.1 |
TC0200012488.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200191122.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191122.hg.1 |
NR_104182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012488.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012488.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012488.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012488.hg.1 |
ENST00000506009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012488.hg.1 |
NR_104182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012488.hg.1 |
NR_104182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012488.hg.1 |
NR_104182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012488.hg.1 |
NR_104182.1 |
Refseq |