Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000433001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000433001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000433001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000433001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
NR_103857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
NR_103857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
NR_103857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
NR_103857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012429.hg.1 |
NR_103857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000433001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000433001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000433001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000433001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000451140.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
ENST00000585280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
NR_103857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
NR_103857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
NR_103857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
NR_103857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012429.hg.1 |
NR_103857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206711.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206711.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206711.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206711.hg.1 |
TC1700012429.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700206711.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206711.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206711.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206712.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206712.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206712.hg.1 |
ENST00000433001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206712.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206712.hg.1 |
TC1700012429.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700206712.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206712.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206712.hg.1 |
ENST00000433001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206712.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206713.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206713.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206713.hg.1 |
ENST00000433001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206713.hg.1 |
NR_103856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206713.hg.1 |
TC1700012429.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700206713.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206713.hg.1 |
ENST00000411573.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206713.hg.1 |
ENST00000433001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206713.hg.1 |
NR_103856.1 |
Refseq |