Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010597.hg.1 |
ENST00000546508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010597.hg.1 |
ENST00000546508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010597.hg.1 |
ENST00000546508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010597.hg.1 |
NR_073454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010597.hg.1 |
NR_073454.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010597.hg.1 |
NR_073454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010597.hg.1 |
ENST00000546508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010597.hg.1 |
ENST00000546508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010597.hg.1 |
ENST00000546508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010597.hg.1 |
NR_073454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010597.hg.1 |
NR_073454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010597.hg.1 |
NR_073454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179196.hg.1 |
ENST00000546508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179196.hg.1 |
NR_073454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179196.hg.1 |
TC1400010597.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400179196.hg.1 |
NR_073454.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179196.hg.1 |
ENST00000546508.1 |
Ensembl |