Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008978.hg.1 |
ENST00000520149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008978.hg.1 |
ENST00000520149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008978.hg.1 |
ENST00000520149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008978.hg.1 |
ENST00000520149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008978.hg.1 |
ENST00000520149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008978.hg.1 |
NR_125424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008978.hg.1 |
NR_125424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008978.hg.1 |
NR_125424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008978.hg.1 |
NR_125424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008978.hg.1 |
NR_125424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161275.hg.1 |
ENST00000520149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161275.hg.1 |
NR_125424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161275.hg.1 |
TC0800008978.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800161275.hg.1 |
NR_125424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161275.hg.1 |
ENST00000520149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161274.hg.1 |
ENST00000520149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161274.hg.1 |
NR_125424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161274.hg.1 |
TC0800008978.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800161274.hg.1 |
ENST00000520149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161274.hg.1 |
NR_125424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008978.hg.1 |
ENST00000520149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008978.hg.1 |
ENST00000520149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008978.hg.1 |
ENST00000520149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008978.hg.1 |
ENST00000520149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008978.hg.1 |
ENST00000520149.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008978.hg.1 |
NR_125424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008978.hg.1 |
NR_125424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008978.hg.1 |
NR_125424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008978.hg.1 |
NR_125424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008978.hg.1 |
NR_125424.1 |
Refseq |