Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
NR_125405.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
NR_125405.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
NR_125405.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
NR_125405.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008552.hg.1 |
NR_125405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160876.hg.1 |
ENST00000608346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160876.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160876.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160876.hg.1 |
NR_125405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160876.hg.1 |
TC0300008552.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300160876.hg.1 |
NR_125405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160876.hg.1 |
ENST00000608346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160876.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160876.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000608015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000608346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
NR_125405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
TC0300008552.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000608015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000608346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160868.hg.1 |
NR_125405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000608756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
ENST00000609469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
NR_125405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
NR_125405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
NR_125405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
NR_125405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008552.hg.1 |
NR_125405.1 |
Refseq |