Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
TC0200007792.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153814.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
TC0200007792.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153818.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |