Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007005.hg.1 |
ENST00000447570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007005.hg.1 |
ENST00000447570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007005.hg.1 |
ENST00000452690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007005.hg.1 |
ENST00000452690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007005.hg.1 |
ENST00000452690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007005.hg.1 |
ENST00000452690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007005.hg.1 |
NR_110386.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007005.hg.1 |
NR_110386.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007005.hg.1 |
NR_110386.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007005.hg.1 |
NR_110386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147639.hg.1 |
ENST00000447570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147639.hg.1 |
ENST00000452690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147639.hg.1 |
NR_110386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147639.hg.1 |
TC0X00007005.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00147639.hg.1 |
ENST00000447570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147639.hg.1 |
ENST00000452690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147639.hg.1 |
NR_110386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147642.hg.1 |
ENST00000452690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147642.hg.1 |
NR_110386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147642.hg.1 |
TC0X00007005.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00147642.hg.1 |
ENST00000452690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147642.hg.1 |
NR_110386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007005.hg.1 |
ENST00000447570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007005.hg.1 |
ENST00000447570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007005.hg.1 |
ENST00000452690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007005.hg.1 |
ENST00000452690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007005.hg.1 |
ENST00000452690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007005.hg.1 |
ENST00000452690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007005.hg.1 |
NR_110386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007005.hg.1 |
NR_110386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007005.hg.1 |
NR_110386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007005.hg.1 |
NR_110386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147638.hg.1 |
ENST00000452690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147638.hg.1 |
NR_110386.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147638.hg.1 |
TC0X00007005.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00147638.hg.1 |
ENST00000452690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147638.hg.1 |
NR_110386.1 |
Refseq |