Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
ENST00000416607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
NR_110256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
NR_110256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
NR_110256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
NR_110256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
NR_110256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
NR_110256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011649.hg.1 |
NR_110256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184573.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184573.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184573.hg.1 |
NR_110256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184573.hg.1 |
TC0200011649.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200184573.hg.1 |
NR_110256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200184573.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200184573.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
ENST00000416607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
NR_110256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
NR_110256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
NR_110256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
NR_110256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
NR_110256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
NR_110256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011649.hg.1 |
NR_110256.1 |
Refseq |