Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04403524_m1
          See other LOC100506682 GE Assays ›
          Gene Symbol:
          LOC100506682
          Gene Name:
          uncharacterized LOC100506682
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 127476883 - 127485804 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04403524_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100506682
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_110024.1 1 - 2 93 120
          GenBank mRNA BM715667.1 - 1 - 2 93 120
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BM701167.1 - 1 - 2 93 Chr. 7 - 127476883 - 127485207 [+] on Build GRCh38 120
          GenBankmRNA BM676233.1 - 3 - 4 Chr. 7 - 127476883 - 127485302 [-] on Build GRCh38 120

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger protein 800 ZNF800 -84.2kb Anti-sense Chr. 7 - 127342871 - 127392803 on NCBI Build GRCh38
          Downstream GRIP and coiled-coil domain containing 1 GCC1 GCC1P,
          GCC88
          103.0kb Anti-sense Chr. 7 - 127580628 - 127585600 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700009016.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0700009016.hg.1 ENST00000448311.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009016.hg.1 ENST00000448311.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009016.hg.1 ENST00000448311.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009016.hg.1 ENST00000448311.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009016.hg.1 NR_110024.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009016.hg.1 NR_110024.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009016.hg.1 NR_110024.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009016.hg.1 NR_110024.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700163900.hg.1 ENST00000448311.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700163900.hg.1 NR_110024.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700163900.hg.1 TC0700009016.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700163900.hg.1 NR_110024.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700163900.hg.1 ENST00000448311.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009016.hg.1 ENST00000448311.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009016.hg.1 ENST00000448311.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009016.hg.1 ENST00000448311.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009016.hg.1 ENST00000448311.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009016.hg.1 NR_110024.1 Refseq
          Affymetrix Human Clariom D Assay TC0700009016.hg.1 NR_110024.1 Refseq
          Affymetrix Human Clariom D Assay TC0700009016.hg.1 NR_110024.1 Refseq
          Affymetrix Human Clariom D Assay TC0700009016.hg.1 NR_110024.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700163899.hg.1 ENST00000448311.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700163899.hg.1 NR_110024.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700163899.hg.1 TC0700009016.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700163899.hg.1 ENST00000448311.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700163899.hg.1 NR_110024.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5db945b66b-v5hlb:80/100.66.134.185:80.
          git-commit: 84360441031e17f3391aad9879d55c303cd12901
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0