Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008698.hg.1 |
ENST00000629221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008698.hg.1 |
ENST00000629221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008698.hg.1 |
ENST00000629221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008698.hg.1 |
NR_109813.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008698.hg.1 |
NR_109813.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008698.hg.1 |
NR_109813.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008698.hg.1 |
ENST00000629221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008698.hg.1 |
ENST00000629221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008698.hg.1 |
ENST00000629221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008698.hg.1 |
NR_109813.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008698.hg.1 |
NR_109813.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008698.hg.1 |
NR_109813.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158931.hg.1 |
ENST00000629221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158931.hg.1 |
NR_109813.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158931.hg.1 |
TC0X00008698.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00158931.hg.1 |
NR_109813.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158931.hg.1 |
ENST00000629221.1 |
Ensembl |