Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04399392_m1
          See other ATP6AP1L GE Assays ›
          Gene Symbol:
          ATP6AP1L
          Gene Name:
          ATPase H+ transporting accessory protein 1 like
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 82278645 - 82344294 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04399392_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ATP6AP1L
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017971.2 NP_001017971.1 8 - 9 1447 92
          XM_011543719.2 XP_011542021.1 7 - 8 1534 92
          XM_011543720.2 XP_011542022.1 6 - 7 1457 92
          XM_011543723.2 XP_011542025.1 8 - 9 2094 92
          XM_017010025.1 XP_016865514.1 7 - 8 1180 92
          XM_017010026.1 XP_016865515.1 8 - 9 1637 92
          XM_017010027.1 XP_016865516.1 8 - 9 985 92
          XM_017010028.1 XP_016865517.1 8 - 9 1270 92
          XM_017010029.1 XP_016865518.1 7 - 8 1586 92
          XM_017010030.1 XP_016865519.1 7 - 8 1219 92
          XM_017010031.1 XP_016865520.1 8 - 9 1625 92
          XM_017010032.1 XP_016865521.1 9 - 10 1795 92
          XM_017010033.1 XP_016865522.1 8 - 9 1258 92
          XM_017010034.1 XP_016865523.1 7 - 8 1583 92
          XM_017010035.1 XP_016865524.1 6 - 7 1143 92
          XM_017010036.1 XP_016865525.1 7 - 8 1558 92
          XM_017010037.1 XP_016865526.1 6 - 7 1495 92
          XM_017010038.1 XP_016865527.1 7 - 8 1539 92
          XM_017010039.1 XP_016865528.1 3 - 4 253 92
          XM_017010040.1 XP_016865529.1 3 - 4 158 92
          XM_017010041.1 XP_016865530.1 9 - 10 2172 92
          XM_017010042.1 XP_016865531.1 10 - 11 2078 92
          XM_017010043.1 XP_016865532.1 9 - 10 2161 92
          XM_017010044.1 XP_016865533.1 8 - 9 2120 92
          XM_017010045.1 XP_016865534.1 8 - 9 1672 92
          XM_017010046.1 XP_016865535.1 8 - 9 1753 92
          XM_017010047.1 XP_016865536.1 7 - 8 2042 92
          GenBank mRNA AK022625.1 - 8 - 9 1447 92
          BC093982.1 - 2 - 3 177 7939827 92
          BC112191.1 - 2 - 3 177 8327659 92

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ATP6AP1L

          Gene Ontology Categories:

          Process(es)

          proton-transporting ATP synthase activity, rotational mechanism
          proton-transporting ATPase activity, rotational mechanism

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0