Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020040.hg.1 |
NR_045006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020040.hg.1 |
TC09001594.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020041.hg.1 |
ENST00000517270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020041.hg.1 |
NR_045006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020041.hg.1 |
TC09001594.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020042.hg.1 |
NR_045006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020042.hg.1 |
TC09001594.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0900178750.hg.1 |
NR_045006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178750.hg.1 |
TC0900011548.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900178750.hg.1 |
NR_045006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178751.hg.1 |
ENST00000517270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178751.hg.1 |
NR_045006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178751.hg.1 |
TC0900011548.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900178751.hg.1 |
ENST00000517270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178751.hg.1 |
NR_045006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178752.hg.1 |
NR_045006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178752.hg.1 |
TC0900011548.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900178752.hg.1 |
NR_045006.1 |
Refseq |