Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04273891_m1
          See other LOC401177 GE Assays ›
          Gene Symbol:
          LOC401177
          Gene Name:
          uncharacterized LOC401177
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 17378906 - 17387310 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04273891_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC401177
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033975.1 2 - 3 709 115
          GenBank mRNA BC052942.1 - 2 - 3 709 115

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream brain abundant membrane attached signal protein 1 BASP1 CAP-23,
          CAP23,
          NAP-22,
          NAP22
          -102.1kb Anti-sense Chr. 5 - 17216823 - 17276845 on NCBI Build GRCh38
          Downstream uncharacterized LOC101929544 LOC101929544 18.7kb Anti-sense Chr. 5 - 17404019 - 17441694 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500010221.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05001191.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961162 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961243 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794202 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961151 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794136 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794130 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794153 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794218 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961181 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961218 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794166 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794172 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961165 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794158 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15668752 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794122 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794123 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961178 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794139 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794163 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794125 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794140 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794214 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794118 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794129 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15668753 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794167 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961200 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961241 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961197 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961166 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961257 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961172 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961198 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961201 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961145 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961237 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961156 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961152 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794243 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794146 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961155 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961176 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961242 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961146 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794200 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794204 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794225 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961251 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15668755 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961144 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961169 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15668751 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794141 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961140 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23961183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03794157 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 178205 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 10145 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 160496 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3440240 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 50903 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01188523 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3440240 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3440240 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01188523 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01188523 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0500010221.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010221.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010221.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010221.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010221.hg.1 NR_033975.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010221.hg.1 NR_033975.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010221.hg.1 NR_033975.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010221.hg.1 NR_033975.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05016988.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05016988.hg.1 NR_033975.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05016988.hg.1 TC05001191.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05016990.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05016990.hg.1 NR_033975.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05016990.hg.1 TC05001191.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05016989.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05016989.hg.1 NR_033975.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05016989.hg.1 TC05001191.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0500171522.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171522.hg.1 NR_033975.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171522.hg.1 TC0500010221.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500171522.hg.1 NR_033975.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171522.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010221.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010221.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010221.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010221.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010221.hg.1 NR_033975.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010221.hg.1 NR_033975.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010221.hg.1 NR_033975.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010221.hg.1 NR_033975.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171523.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171523.hg.1 NR_033975.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171523.hg.1 TC0500010221.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500171523.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171523.hg.1 NR_033975.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171525.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171525.hg.1 NR_033975.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171525.hg.1 TC0500010221.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500171525.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171525.hg.1 NR_033975.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171524.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171524.hg.1 NR_033975.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171524.hg.1 TC0500010221.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500171524.hg.1 ENST00000505158.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171524.hg.1 NR_033975.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A026572 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A026572 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0