Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011164.hg.1 |
ENST00000523996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011164.hg.1 |
NR_036611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011164.hg.1 |
NR_036611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011164.hg.1 |
NR_036611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018770.hg.1 |
NR_036611.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018770.hg.1 |
TC08001447.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0800176041.hg.1 |
NR_036611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176041.hg.1 |
TC0800011164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800176041.hg.1 |
NR_036611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176042.hg.1 |
ENST00000523996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176042.hg.1 |
NR_036611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176042.hg.1 |
TC0800011164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800176042.hg.1 |
ENST00000523996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176042.hg.1 |
NR_036611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011164.hg.1 |
ENST00000523996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011164.hg.1 |
NR_036611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011164.hg.1 |
NR_036611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011164.hg.1 |
NR_036611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176043.hg.1 |
NR_036611.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176043.hg.1 |
TC0800011164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800176043.hg.1 |
NR_036611.1 |
Refseq |