Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007827.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007827.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007827.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007827.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007827.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007827.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007827.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004351.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004351.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004351.hg.1 |
TC09000405.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006505.hg.1 |
ENST00000613133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006505.hg.1 |
ENST00000613133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006505.hg.1 |
ENST00000613133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006505.hg.1 |
ENST00000613133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007827.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007827.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007827.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007827.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007827.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007827.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007827.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007827.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900152842.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900152842.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900152842.hg.1 |
TC0900007827.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900152842.hg.1 |
NM_178828.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900152842.hg.1 |
ENST00000325643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006505.hg.1 |
ENST00000613133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006505.hg.1 |
ENST00000613133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006505.hg.1 |
ENST00000613133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006505.hg.1 |
ENST00000613133.1 |
Ensembl |