Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013352.hg.1 |
ENST00000331459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013352.hg.1 |
NM_002548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013352.hg.1 |
TC17001002.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008668.hg.1 |
ENST00000448731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008668.hg.1 |
ENST00000448731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009470.hg.1 |
ENST00000331459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009470.hg.1 |
NM_002548.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006579.hg.1 |
ENST00000573526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009560.hg.1 |
ENST00000589199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009470.hg.1 |
ENST00000331459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009470.hg.1 |
NM_002548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006579.hg.1 |
ENST00000573526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008668.hg.1 |
ENST00000448731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008668.hg.1 |
ENST00000448731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009560.hg.1 |
ENST00000589199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700172417.hg.1 |
ENST00000331459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700172417.hg.1 |
NM_002548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700172417.hg.1 |
TC1700009470.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700172417.hg.1 |
NM_002548.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700172417.hg.1 |
ENST00000331459.1 |
Ensembl |