Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03923202_s1
          See other P2RY1 GE Assays ›
          Gene Symbol:
          P2RY1
          Gene Name:
          purinergic receptor P2Y1
          Gene Aliases:
          P2Y1
          Chromosome Location:
          Chr.3: 152834693 - 152841439 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03923202_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: P2RY1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002563.4 NP_002554.1 1 5148 78
          GenBank mRNA AK026659.1 - 1 451 78
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA127714.1 - 1 151 Chr. 3 - 152839690 - 152840173 [+] on Build GRCh38 78
          GenBankmRNA DB256942.1 - 1 354 Chr. 3 - 152839487 - 152840059 [+] on Build GRCh38 78
          GenBankmRNA AA130371.1 - 1 151 Chr. 3 - 152839690 - 152839971 [+] on Build GRCh38 78
          GenBankmRNA DA570315.1 - 1 47 Chr. 3 - 152839794 - 152840524 [+] on Build GRCh38 78
          GenBankmRNA AA403227.1 - 1 143 Chr. 3 - 152839698 - 152840254 [+] on Build GRCh38 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Downstream RAP2B,
          member of RAS oncogene family
          RAP2B 320.8kb Sense Chr. 3 - 153162212 - 153170624 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene P2RY1

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          G protein-coupled adenosine receptor signaling pathway
          cell surface receptor signaling pathway
          G protein-coupled receptor signaling pathway
          adenylate cyclase-inhibiting G protein-coupled receptor signaling pathway
          phospholipase C-activating G protein-coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          aging
          glial cell migration
          response to mechanical stimulus
          regulation of signaling receptor activity
          negative regulation of norepinephrine secretion
          sensory perception of pain
          signal transduction involved in regulation of gene expression
          platelet activation
          positive regulation of inositol trisphosphate biosynthetic process
          G protein-coupled purinergic nucleotide receptor signaling pathway
          eating behavior
          positive regulation of ion transport
          positive regulation of transcription by RNA polymerase II
          positive regulation of hormone secretion
          negative regulation of binding
          positive regulation of penile erection
          positive regulation of ERK1 and ERK2 cascade
          response to growth factor
          cellular response to organic cyclic compound
          protein localization to plasma membrane
          relaxation of muscle
          blood vessel diameter maintenance
          G protein-coupled ADP receptor activity
          protein binding
          ATP binding
          A1 adenosine receptor binding
          signaling receptor activity
          ADP binding
          G protein-coupled purinergic nucleotide receptor activity
          G protein-coupled ATP receptor activity
          protein heterodimerization activity
          scaffold protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0300009224.hg.1 ENST00000305097.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009224.hg.1 ENST00000305097.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009224.hg.1 ENST00000305097.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009224.hg.1 ENST00000305097.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009224.hg.1 ENST00000305097.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009224.hg.1 ENST00000305097.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009224.hg.1 ENST00000460407.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009224.hg.1 NM_002563.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009224.hg.1 NM_002563.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009224.hg.1 NM_002563.4 Refseq
          Affymetrix Human Clariom D Assay TC0300009224.hg.1 ENST00000305097.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009224.hg.1 ENST00000305097.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009224.hg.1 ENST00000305097.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009224.hg.1 ENST00000305097.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009224.hg.1 ENST00000305097.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009224.hg.1 ENST00000305097.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009224.hg.1 ENST00000460407.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009224.hg.1 NM_002563.4 Refseq
          Affymetrix Human Clariom D Assay TC0300009224.hg.1 NM_002563.4 Refseq
          Affymetrix Human Clariom D Assay TC0300009224.hg.1 NM_002563.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 203228 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0