Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03912953_s1
          See other HS.496185-PLEKHC1 FUSION GE Assays ›
          Gene Symbol:
          Hs.496185-PLEKHC1 fusion KLF8
          Gene Name:
          FUSION
          Kruppel like factor 8
          Gene Aliases:
          Hs.496185, PLEKHC1
          BKLF3, ZNF741
          Chromosome Location:
          -
          Chromosome Location:
          Chr.X: 55908123 - 56291531 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs03912953_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hs.496185-PLEKHC1 fusion
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK091532.1 - 543 111
          Gene Symbol: KLF8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001324105.1 NP_001311034.1 1 543 111
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CB243528.1 - 3 Chr. X - 55908306 - 56205093 [-] on Build GRCh38 111
          GenBankmRNA BU730964.1 - 3 Chr. X - 55908515 - 56205093 [-] on Build GRCh38 111
          GenBankmRNA CD387915.1 - 464 Chr. - - [-] on Build 111
          GenBankmRNA CF130112.1 - 1 285 Chr. X - 55908381 - 56015147 [+] on Build GRCh38 111
          GenBankmRNA BM708630.1 - 1 157 Chr. X - 55908509 - 56205093 [+] on Build GRCh38 111
          GenBankmRNA CA440080.1 - 2 Chr. X - 55908478 - 56015178 [-] on Build GRCh38 111
          GenBankmRNA AI742003.1 - 3 Chr. X - 55908533 - 56205093 [-] on Build GRCh38 111
          GenBankmRNA BX109741.1 - 1 199 Chr. X - 55908467 - 56014961 [+] on Build GRCh38 111

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream Ras related GTP binding B RRAGB RAGB,
          bA465E19.1
          -148.8kb Sense Chr. X - 55717677 - 55760209 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_001324105.1 transcripts.
          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0X00011295.hg.1 ENST00000452532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011295.hg.1 ENST00000452532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011295.hg.1 ENST00000452532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011295.hg.1 ENST00000452532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011295.hg.1 ENST00000452532.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011295.hg.1 ENST00000452532.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011295.hg.1 ENST00000452532.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011295.hg.1 ENST00000452532.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011295.hg.1 ENST00000452532.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011295.hg.1 ENST00000452532.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-wxxj5:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0