Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03871572_s1
          See other LOC100289361 GE Assays ›
          Gene Symbol:
          LOC100289361
          Gene Name:
          uncharacterized LOC100289361
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 143000845 - 143001467 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03871572_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100289361
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_034032.1 1 439 56
          GenBank mRNA BC040051.1 - 1 448 56
          BQ017937.1 - 1 56
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI763247.1 - 1 Chr. 3 - 143000957 - 143001380 [+] on Build GRCh38 56
          GenBankmRNA AI824178.1 - 1 Chr. 3 - 143000912 - 143001233 [+] on Build GRCh38 56
          GenBankmRNA BE501699.1 - 1 Chr. 3 - 143000907 - 143001360 [+] on Build GRCh38 56
          GenBankmRNA AI266564.1 - 1 Chr. 3 - 143000907 - 143001269 [+] on Build GRCh38 56
          GenBankmRNA AW614613.1 - 1 Chr. 3 - 143000957 - 143001380 [+] on Build GRCh38 56
          GenBankmRNA AW772201.1 - 1 Chr. 3 - 143000957 - 143001354 [+] on Build GRCh38 56
          GenBankmRNA BE501973.1 - 1 Chr. 3 - 143000865 - 143001380 [+] on Build GRCh38 56
          GenBankmRNA AI675224.1 - 1 Chr. 3 - 143000957 - 143001259 [+] on Build GRCh38 56
          GenBankmRNA BE221807.1 - 1 Chr. 3 - 143000957 - 143001305 [+] on Build GRCh38 56
          GenBankmRNA AW236927.1 - 1 Chr. 3 - 143000908 - 143001377 [+] on Build GRCh38 56
          GenBankmRNA AA778307.1 - 1 Chr. 3 - 143000964 - 143001340 [+] on Build GRCh38 56
          GenBankmRNA BF593232.1 - 1 Chr. 3 - 143000958 - 143001408 [+] on Build GRCh38 56
          GenBankmRNA AI347533.1 - 1 Chr. 3 - 143000907 - 143001344 [+] on Build GRCh38 56
          GenBankmRNA AW299782.1 - 1 Chr. 3 - 143000907 - 143001362 [+] on Build GRCh38 56
          GenBankmRNA AW136045.1 - 1 Chr. 3 - 143000957 - 143001380 [+] on Build GRCh38 56
          GenBankmRNA AI623168.1 - 1 Chr. 3 - 143000908 - 143001352 [+] on Build GRCh38 56
          GenBankmRNA AI627506.1 - 1 Chr. 3 - 143000915 - 143001425 [+] on Build GRCh38 56
          GenBankmRNA AI927189.1 - 1 Chr. 3 - 143000957 - 143001380 [+] on Build GRCh38 56
          GenBankmRNA AW614337.1 - 1 Chr. 3 - 143000958 - 143001404 [+] on Build GRCh38 56
          GenBankmRNA AA969417.1 - 1 Chr. 3 - 143000907 - 143001342 [+] on Build GRCh38 56
          GenBankmRNA AI299571.1 - 1 Chr. 3 - 143000907 - 143001337 [+] on Build GRCh38 56
          GenBankmRNA AI334622.1 - 1 Chr. 3 - 143000957 - 143001314 [+] on Build GRCh38 56
          GenBankmRNA CN364830.1 - 1 373 Chr. 3 - 143000907 - 143001389 [-] on Build GRCh38 56
          GenBankmRNA AW207242.1 - 1 Chr. 3 - 143000957 - 143001340 [+] on Build GRCh38 56
          GenBankmRNA BQ233226.1 - 1 446 Chr. 3 - 143000907 - 143001465 [-] on Build GRCh38 56
          GenBankmRNA AI916830.1 - 1 Chr. 3 - 143000908 - 143001408 [+] on Build GRCh38 56
          GenBankmRNA AI770178.1 - 1 Chr. 3 - 143000907 - 143001419 [+] on Build GRCh38 56
          GenBankmRNA BE218358.1 - 1 Chr. 3 - 143000957 - 143001362 [+] on Build GRCh38 56
          GenBankmRNA BF056291.1 - 1 Chr. 3 - 143000908 - 143001353 [+] on Build GRCh38 56
          GenBankmRNA BE464269.1 - 1 Chr. 3 - 143000957 - 143001405 [+] on Build GRCh38 56
          GenBankmRNA AA428643.1 - 1 Chr. 3 - 143000908 - 143001296 [+] on Build GRCh38 56
          GenBankmRNA AI308144.1 - 1 Chr. 3 - 143000957 - 143001309 [+] on Build GRCh38 56
          GenBankmRNA AW614222.1 - 1 Chr. 3 - 143000957 - 143001380 [+] on Build GRCh38 56
          GenBankmRNA BE672060.1 - 1 Chr. 3 - 143000907 - 143001355 [+] on Build GRCh38 56
          GenBankmRNA AW770482.1 - 1 Chr. 3 - 143000957 - 143001362 [+] on Build GRCh38 56
          GenBankmRNA AW002419.1 - 1 Chr. 3 - 143000907 - 143001335 [+] on Build GRCh38 56

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream PAQR9 antisense RNA 1 PAQR9-AS1 Anti-sense Chr. 3 - 142964058 - 143001090 on NCBI Build GRCh38
          Downstream U2 snRNP associated SURP domain containing U2SURP SR140,
          fSAPa
          57.0 Anti-sense Chr. 3 - 143001524 - 143060725 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300012680.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001861.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P209791 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 9356 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P209791 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P209791 - -
          Agilent Human 1B Microarray G4111A A_32_P209791 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0300012680.hg.1 ENST00000597953.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012680.hg.1 NR_034032.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012680.hg.1 NR_034032.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03033591.hg.1 NR_034032.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03033591.hg.1 TC03001861.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0300194799.hg.1 NR_034032.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300194799.hg.1 TC0300012680.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300194799.hg.1 NR_034032.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300194800.hg.1 ENST00000597953.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300194800.hg.1 NR_034032.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300194800.hg.1 TC0300012680.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300194800.hg.1 ENST00000597953.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300194800.hg.1 NR_034032.1 Refseq
          Affymetrix Human Clariom D Assay TC0300012680.hg.1 ENST00000597953.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012680.hg.1 NR_034032.1 Refseq
          Affymetrix Human Clariom D Assay TC0300012680.hg.1 NR_034032.1 Refseq
          Human Genome U95 Set 89959_PM_at - -
          Human Genome U95 Set 89959_at - -
          Human Genome U133 Set 229338_at - -
          Human Genome U133 Set 229338_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A021703 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A021703 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0