Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927603 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK091880.1 | - | 1 | 1643 | 96 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | DB320479.1 | - | 1 | Chr. 9 - 85536440 - 85536975 [+] on Build GRCh38 | 96 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | neurotrophic receptor tyrosine kinase 2 | NTRK2 |
GP145-TrkB, TRKB, trk-B |
-509.4kb | Anti-sense | Chr. 9 - 84668368 - 85027070 on NCBI Build GRCh38 |
Downstream | ATP/GTP binding protein 1 | AGTPBP1 |
CCP1, NNA1 |
8.2kb | Sense | Chr. 9 - 85543799 - 85742029 on NCBI Build GRCh38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |