Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000559722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000559722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000559722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000560261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000560261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000560261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000560261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000560261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000560919.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000560919.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000560919.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000567415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000258874.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000559722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000559722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000559722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000560261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000560261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000560261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000560261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000560261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000560919.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000560919.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000560919.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000567415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010900.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185581.hg.1 |
ENST00000567415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185581.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185581.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185581.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185581.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185581.hg.1 |
NR_037654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185581.hg.1 |
TC1500010900.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500185581.hg.1 |
ENST00000615374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185581.hg.1 |
ENST00000567415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185581.hg.1 |
ENST00000611007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185581.hg.1 |
NM_001199758.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185581.hg.1 |
NM_006441.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185581.hg.1 |
NR_037654.1 |
Refseq |