Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03681731_s1
          See other LINC01000 GE Assays ›
          Gene Symbol:
          LINC01000
          Gene Name:
          long intergenic non-protein coding RNA 1000
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 128641241 - 128660998 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03681731_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01000
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024368.1 5 9951 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream methyltransferase like 2B METTL2B METL,
          METTL2,
          METTL2A,
          PSENIP1
          -148.3kb Sense Chr. 7 - 128476729 - 128502924 on NCBI Build GRCh38
          Downstream family with sequence similarity 71 member F2 FAM71F2 FAM137B 10.6kb Sense Chr. 7 - 128671628 - 128702262 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26031619 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 114192 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 671791 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 350603 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 476912 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 669088 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 926681 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 536050 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 113757 AJ496730.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 113757 AK022695.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 113757 AL136929.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 113757 AY094972.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 113757 BC051818.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 113757 BC110895.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 113757 NM_001033568.1 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 329565 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 87630 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 143245 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 252678 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 251723 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 115721 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3362397 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3278881 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P41375 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P24741 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P41375 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P50406 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 4459 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 38743 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 39366 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 25072 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 52248 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 26426 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 45620 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P18111905 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3362397 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3369213 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3278881 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P41375 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P583225 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P41375 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3278881 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3362397 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05774118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05774118 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P24741 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P41375 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P50406 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P01804988 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P01804988 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P24741 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P12183 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P50406 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P11230 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P436336 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P41375 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P665504 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P898915 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P583225 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P586390 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P75425 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 98033 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 85900 - -
          Agilent Human 1B Microarray G4111A A_32_P57326 - -
          Agilent Human 1B Microarray G4111A A_32_P24741 - -
          Agilent Human 1B Microarray G4111A A_32_P41375 - -
          Agilent Human 1B Microarray G4111A A_32_P363833 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05774142 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05774118 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16016731 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05774127 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05774123 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05774133 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05774119 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05774131 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05774136 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05774145 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05774135 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05774125 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05774138 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18111905 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8031993 - -
          Affymetrix Human Gene 1.0 ST Array 7945345 - -
          Affymetrix Human Gene 1.0 ST Array 8031995 - -
          Affymetrix Human Gene 1.0 ST Array 8031996 - -
          Affymetrix Human Gene 1.0 ST Array 8133029 - -
          Affymetrix Human Gene 1.0 ST Array 8135932 - -
          Affymetrix Human Gene 1.0 ST Array 8133028 - -
          Affymetrix Human Gene 1.0 ST Array 8031994 - -
          Affymetrix Human Gene 1.1 ST Array 8133027 - -
          Affymetrix Human Gene 1.1 ST Array 7945344 - -
          Affymetrix Human Gene 1.1 ST Array 8031992 - -
          Affymetrix Human Gene 1.1 ST Array 8135931 - -
          Affymetrix HT HG-U133+ PM Array Plate 227129_PM_x_at - -
          Affymetrix HT HG-U133+ PM Array Plate 238678_PM_at - -
          Affymetrix Human Genome U219 Array 11745798_x_at - -
          Affymetrix Human Genome U219 Array 11745798_PM_x_at - -
          Affymetrix Human Genome U219 Array 11745797_PM_x_at - -
          Affymetrix Human Genome U219 Array 11745797_x_at - -
          Human Genome U133 Plus 2.0 Array 231821_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 231821_x_at - -
          Human Genome U133 Plus 2.0 Array 238678_PM_at - -
          Human Genome U133 Plus 2.0 Array 238678_at - -
          Human Genome U133 Plus 2.0 Array 227129_x_at - -
          Human Genome U133 Plus 2.0 Array 225899_x_at - -
          Human Genome U133 Plus 2.0 Array 227129_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 225899_PM_x_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227129_x_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238678_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238678_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227129_x_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238678_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238678_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227129_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238678_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227129_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227129_x_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238678_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238678_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227129_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227129_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227129_x_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227129_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238678_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238678_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227129_x_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238678_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227129_x_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 238678_at5 - -
          Human Genome U95 Set 62705_at - -
          Human Genome U95 Set 65794_PM_f_at - -
          Human Genome U95 Set 65795_PM_r_at - -
          Human Genome U95 Set 65795_r_at - -
          Human Genome U95 Set 65794_f_at - -
          Human Genome U95 Set 62705_PM_at - -
          Human Genome U133 Set 231821_x_at - -
          Human Genome U133 Set 225899_x_at - -
          Human Genome U133 Set 238678_PM_at - -
          Human Genome U133 Set 225899_PM_x_at - -
          Human Genome U133 Set 227129_x_at - -
          Human Genome U133 Set 238678_at - -
          Human Genome U133 Set 231821_PM_x_at - -
          Human Genome U133 Set 227129_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035030 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035033 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035031 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A011562 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035032 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A011562 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035033 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035030 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035031 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035032 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 278671 - -
          ABI Human Genome Survey Microarray Version 2.0 417387 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3249711 NR_024368.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1743569 - -
          HumanHT-12 V4.0 expression beadchip ILMN_3249711 NR_024368.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3249711 NR_024368.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3249711 NR_024368.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1743569 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1699131 AK127273.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1699131 NR_024368.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1699131 AK127273.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1699131 NR_024368.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7d7666c9cf-hbrf6:80/100.66.134.79:80.
          git-commit: 173eb311d069d8bb3c8868db8478582b9fe5b4f6
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0