Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03672620_s1
          See other LOC100130642 GE Assays ›
          Gene Symbol:
          LOC100130642
          Gene Name:
          uncharacterized LOC100130642
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03672620_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100130642
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK124249.1 - 1 2655 84

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream CXXC repeat containing interactor of PDZ3 domain CRIPT HSPC139,
          SSMDF
          -16.3kb Sense Chr. 2 - 46617172 - 46630176 on NCBI Build GRCh38
          Downstream suppressor of cytokine signaling 5 SOCS5 CIS6,
          CISH6,
          Cish5,
          SOCS-5
          49.8kb Sense Chr. 2 - 46698960 - 46762788 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153314 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233804 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153311 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233821 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15240272 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153305 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233779 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233789 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153304 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153296 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153316 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15240279 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153291 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153300 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153297 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153328 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153317 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233803 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153292 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233785 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153323 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233807 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233815 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153335 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15240269 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233806 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153326 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233798 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233797 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153331 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233783 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233816 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153318 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233794 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153333 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15240275 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153320 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233775 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233795 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153319 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21233793 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01153307 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3254535 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 32904 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3254535 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P13195553 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 243970_PM_at - -
          Human Genome U133 Plus 2.0 Array 243970_at - -
          Human Genome U133 Plus 2.0 Array 243970_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243970_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243970_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243970_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243970_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243970_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243970_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243970_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243970_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243970_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243970_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243970_at7 - -
          Human Genome U95 Set 85775_PM_at - -
          Human Genome U95 Set 85775_at - -
          Human Genome U133 Set 243970_PM_at - -
          Human Genome U133 Set 243970_at - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1866289 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 5490685 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_81105 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1866289 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1866289 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1866289 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0