Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000360899.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000360899.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000360899.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000553623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000553623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000554195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000554195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000554195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000556308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000556308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000556308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000556308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000556308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000556697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000556697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000556697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
ENST00000556697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
NR_024096.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
NR_024096.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008222.hg.1 |
NR_024096.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000360899.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000360899.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000360899.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000553367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000553623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000553623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000554195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000554195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000554195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000556308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000556308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000556308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000556308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000556308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000556697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000556697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000556697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
ENST00000556697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
NR_024096.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
NR_024096.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008222.hg.1 |
NR_024096.1 |
Refseq |