Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03669719_s1
          See other TRABD2B GE Assays ›
          Gene Symbol:
          TRABD2B
          Gene Name:
          TraB domain containing 2B
          Gene Aliases:
          TIKI2
          Chromosome Location:
          Chr.1: 47760528 - 47997367 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03669719_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRABD2B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001194986.1 NP_001181915.1 7 6007 119
          XM_006710640.3 XP_006710703.1 8 6588 119
          XM_017001260.1 XP_016856749.1 7 6318 119
          XM_017001261.1 XP_016856750.1 5 6038 119
          XM_017001263.1 XP_016856752.1 5 6457 119
          GenBank mRNA AK090467.1 - 1 3447 119

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream forkhead box D2 FOXD2 FKHL17,
          FREAC-9,
          FREAC9
          -319.8kb Anti-sense Chr. 1 - 47436017 - 47440691 on NCBI Build GRCh38
          Downstream uncharacterized LOC105378702 LOC105378702 50.9kb Anti-sense Chr. 1 - 47815880 - 47819491 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100014109.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002634.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          Wnt signaling pathway
          negative regulation of Wnt signaling pathway
          positive regulation of protein oligomerization
          positive regulation of protein oxidation
          metalloendopeptidase activity
          protein binding
          Wnt-protein binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01041190.hg.1 ENST00000438589.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01041190.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01041190.hg.1 TC01002634.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000438589.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000438589.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000435576.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000435576.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000435576.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014109.hg.1 ENST00000435576.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01041189.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01041189.hg.1 TC01002634.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100205577.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100205577.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100205577.hg.1 TC0100014109.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100205577.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100205577.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100205579.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100205579.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100205579.hg.1 TC0100014109.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100205579.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100205579.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000438589.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000438589.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000435576.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000435576.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000435576.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100014109.hg.1 ENST00000435576.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100205580.hg.1 ENST00000438589.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100205580.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100205580.hg.1 NM_001194986.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100205580.hg.1 TC0100014109.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100205580.hg.1 ENST00000438589.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100205580.hg.1 ENST00000606738.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100205580.hg.1 NM_001194986.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 151381 - -
          ABI Human Genome Survey Microarray Version 2.0 152827 - -
          ABI Human Genome Survey Microarray Version 2.0 208917 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0