Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03665212_m1
          See other TISP43 GE Assays ›
          Gene Symbol:
          TISP43 LOC646743
          Gene Name:
          uncharacterized LOC150527
          uncharacterized LOC646743
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.2: 130570846 - 130584161 on Build GRCh38
          Chromosome Location:
          Chr.2: 130536570 - 130549915 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs03665212_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TISP43
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027313.1 2 - 3 309 88
          GenBank mRNA BC064385.1 - 2 - 3 309 88
          Gene Symbol: LOC646743
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033930.1 2 - 3 326 88
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BM559529.1 - 2 - 3 322 Chr. 2 - 130570847 - 130582871 [+] on Build GRCh38 88

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cripto,
          FRL-1,
          cryptic family 1B
          CFC1B -8.6kb Anti-sense Chr. 2 - 130521094 - 130527992 on NCBI Build GRCh38
          Downstream cripto,
          FRL-1,
          cryptic family 1
          CFC1 CFC1B,
          CRYPTIC,
          DTGA2,
          HTX2
          8.0kb Anti-sense Chr. 2 - 130592165 - 130599575 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200014273.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000815.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 432819 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 585114 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 293151 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 904818 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 687187 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 373177 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 766536 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 260699 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 658523 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 888411 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 478805 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 184582 BC107048.2 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 184582 NM_178491.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 257835 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 237741 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3242632 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3403494 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3221403 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 13078 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 52400 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 49037 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3242632 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3403494 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3403494 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3242632 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3403494 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8055237 - -
          Affymetrix Human Gene 1.0 ST Array 8045207 - -
          Affymetrix Human Gene 1.0 ST Array 8055238 - -
          Affymetrix Human Gene 1.0 ST Array 8045207 - -
          Affymetrix Human Gene 1.0 ST Array 8045206 - -
          Affymetrix Human Gene 1.0 ST Array 8055237 - -
          Affymetrix Human Gene 1.1 ST Array 8055236 - -
          Affymetrix Human Gene 1.1 ST Array 8045205 - -
          Affymetrix Human Gene 1.1 ST Array 8045205 - -
          Affymetrix Human Gene 1.1 ST Array 8055236 - -
          Affymetrix HT HG-U133+ PM Array Plate 1560806_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR02012556.hg.1 ENST00000409793.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02012556.hg.1 NR_027313.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02012556.hg.1 NR_027314.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02012556.hg.1 NR_033930.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02012556.hg.1 TC02000815.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000409793.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000409793.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000409793.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000409793.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000409793.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000409982.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000409982.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000409982.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000409982.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000409982.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000414595.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000414595.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000414595.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000414595.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000414595.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000419965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000419965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000419965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000419965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000419965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000419965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000419965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000440359.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000440359.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000440359.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000440359.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000623376.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000623376.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000623376.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000623376.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000623376.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000623376.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009307.hg.1 ENST00000623376.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014273.hg.1 ENST00000409259.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014273.hg.1 ENST00000409259.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014273.hg.1 ENST00000409259.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014273.hg.1 ENST00000409259.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014273.hg.1 ENST00000409259.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014273.hg.1 NR_027313.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014273.hg.1 NR_027313.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014273.hg.1 NR_027313.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014273.hg.1 NR_027314.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014273.hg.1 NR_027314.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014273.hg.1 NR_033930.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014273.hg.1 NR_033930.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014273.hg.1 NR_033930.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02012551.hg.1 ENST00000409982.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02012551.hg.1 NR_027313.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02012551.hg.1 NR_033930.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02012551.hg.1 TC02000815.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000409793.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000409793.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000409793.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000409793.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000409793.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000409982.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000409982.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000409982.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000409982.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000409982.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000414595.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000414595.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000414595.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000414595.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000414595.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000419965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000419965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000419965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000419965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000419965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000419965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000419965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000440359.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000440359.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000440359.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000440359.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000623376.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000623376.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000623376.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000623376.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000623376.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000623376.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200009307.hg.1 ENST00000623376.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200204308.hg.1 NR_027313.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200204308.hg.1 NR_027314.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200204308.hg.1 NR_033930.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200204308.hg.1 TC0200014273.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200204308.hg.1 NR_033930.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200204308.hg.1 NR_027313.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200204308.hg.1 NR_027314.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014273.hg.1 ENST00000409259.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014273.hg.1 ENST00000409259.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014273.hg.1 ENST00000409259.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014273.hg.1 ENST00000409259.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014273.hg.1 ENST00000409259.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014273.hg.1 NR_027313.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014273.hg.1 NR_027313.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014273.hg.1 NR_027313.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014273.hg.1 NR_027314.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014273.hg.1 NR_027314.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014273.hg.1 NR_033930.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014273.hg.1 NR_033930.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014273.hg.1 NR_033930.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200204312.hg.1 ENST00000409259.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200204312.hg.1 NR_027313.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200204312.hg.1 NR_033930.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200204312.hg.1 TC0200014273.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200204312.hg.1 ENST00000409259.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200204312.hg.1 NR_027313.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200204312.hg.1 NR_033930.1 Refseq
          Human Genome U133 Plus 2.0 Array 1560806_PM_at - -
          Human Genome U133 Plus 2.0 Array 1560806_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560806_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560806_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560806_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560806_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560806_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560806_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560806_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560806_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560806_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560806_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560806_at9 - -
          Human Genome U95 Set 66652_PM_at - -
          Human Genome U95 Set 66652_at - -
          Human Genome U95 Set 52216_at - -
          Human Genome U95 Set 52216_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007501 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014350 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014350 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007501 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014350 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014350 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1734775 AY726562.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1734775 AY726562.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1734775 AY726562.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1734775 AY726562.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1734775 AY726562.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0