Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03664654_s1
          See other LOC100034248 GE Assays ›
          Gene Symbol:
          LOC100034248
          Gene Name:
          uncharacterized LOC100034248
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03664654_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100034248
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK128820.1 - 1 1450 87
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA HY213989.1 - 1 Chr. 19 - 7012618 - 7013038 [-] on Build GRCh38 87

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microtubule affinity regulating kinase 1 pseudogene FLJ25758 -5.1kb Anti-sense Chr. 19 - 7004953 - 7006415 on NCBI Build GRCh38
          Downstream methyl-CpG binding domain protein 3 like 5 MBD3L5 16.6kb Sense Chr. 19 - 7030583 - 7033011 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31483599 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17103551 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31483595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31483594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31483578 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31483588 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10871715 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10871723 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31483600 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31483575 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10871719 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10871724 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31483576 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10871718 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10871720 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 313653 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3248973 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 20179 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3248973 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8025125 - -
          Affymetrix Human Gene 1.1 ST Array 8025124 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3259652 AK128820.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3259652 NR_024372.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3259652 AK098624.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3259652 AY090781.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3259652 AK128820.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3259652 NR_024372.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3259652 AK098624.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3259652 AY090781.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-98ffbb479-9qks8:80/100.66.134.79:80.
          git-commit: cc4aab6ff2cd04db3b50ee2b2ac7bb13b20eb648
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0