Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03664003_s1
          See other LOC100129240 GE Assays ›
          Gene Symbol:
          LOC100129240
          Gene Name:
          uncharacterized LOC100129240
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03664003_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100129240
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK128083.1 - 1 3750 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 4455 MIR4455 -45.8kb Sense Chr. 4 - 184938383 - 184938440 on NCBI Build GRCh38
          Downstream helt bHLH transcription factor HELT HCM1228,
          HESL,
          Mgn,
          bHLHb44
          30.7kb Anti-sense Chr. 4 - 185018841 - 185021265 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15646045 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685330 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685344 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685349 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685352 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685334 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685378 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685393 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685377 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685302 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685317 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685296 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846620 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846638 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685382 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685292 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685303 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685379 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846609 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685318 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685321 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685361 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685332 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685339 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846587 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685306 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846628 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846623 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685322 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15646043 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685327 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685331 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685353 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15646048 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685354 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685316 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846550 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685325 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846622 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685304 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685333 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685288 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846579 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846615 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685314 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846619 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685388 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685347 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15646052 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685365 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15646061 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685337 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846614 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846616 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685311 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685309 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685305 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685385 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846532 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846544 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846641 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685294 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685380 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685372 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15646047 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846637 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685291 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685355 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23846624 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03685371 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 195031 AL133023.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 195031 BC033852.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 195031 BC060889.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 195031 BC109035.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 130418 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3253852 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 32545 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01155628 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0400012629.hg.1 ENST00000623949.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012629.hg.1 ENST00000623949.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0