Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000596066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000596066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000596066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000596066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000598170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000598170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000598170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000598170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000598170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
ENST00000598170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
NR_027003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
NR_027003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
NR_027003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
NR_027003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
NR_027003.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008364.hg.1 |
NR_027003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000593980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000596066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000596066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000596066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000596066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000598170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000598170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000598170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000598170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000598170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
ENST00000598170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
NR_027003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
NR_027003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
NR_027003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
NR_027003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
NR_027003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008364.hg.1 |
NR_027003.1 |
Refseq |