Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006812.hg.1 |
ENST00000580967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006812.hg.1 |
ENST00000580967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006812.hg.1 |
ENST00000584531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006812.hg.1 |
ENST00000584531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006812.hg.1 |
ENST00000584531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006812.hg.1 |
ENST00000584531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006812.hg.1 |
NR_104164.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006812.hg.1 |
NR_104164.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006812.hg.1 |
NR_104164.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006812.hg.1 |
NR_104164.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006812.hg.1 |
ENST00000580967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006812.hg.1 |
ENST00000580967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006812.hg.1 |
ENST00000584531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006812.hg.1 |
ENST00000584531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006812.hg.1 |
ENST00000584531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006812.hg.1 |
ENST00000584531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006812.hg.1 |
NR_104164.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006812.hg.1 |
NR_104164.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006812.hg.1 |
NR_104164.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006812.hg.1 |
NR_104164.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800146673.hg.1 |
ENST00000584531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800146673.hg.1 |
NR_104164.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800146673.hg.1 |
TC1800006812.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800146673.hg.1 |
NR_104164.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800146673.hg.1 |
ENST00000584531.1 |
Ensembl |