Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03658712_s1
          See other LOC100132207 GE Assays ›
          Gene Symbol:
          LOC100132207
          Gene Name:
          uncharacterized LOC100132207
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03658712_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100132207
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK123339.1 - 1 1849 91
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BE504172.1 - 1 Chr. 3 - 129609598 - 129610109 [-] on Build GRCh38 91
          GenBankmRNA BQ011337.1 - 1 Chr. 3 - 129609492 - 129610109 [-] on Build GRCh38 91
          GenBankmRNA N32818.1 - 1 Chr. 3 - 129609500 - 129610105 [-] on Build GRCh38 91
          GenBankmRNA BX115968.1 - 1 513 Chr. 3 - 129609146 - 129609896 [+] on Build GRCh38 91

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream plexin D1 PLXND1 PLEXD1 -1.1kb Anti-sense Chr. 3 - 129555213 - 129606739 on NCBI Build GRCh38
          Downstream transmembrane and coiled-coil domain family 1 TMCC1 37.7kb Anti-sense Chr. 3 - 129647792 - 129893605 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3388745 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 46014 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3388745 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P31105940 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P31105940 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 243019_PM_at - -
          Human Genome U133 Plus 2.0 Array 243019_at - -
          Human Genome U133 Plus 2.0 Array 243019_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243019_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243019_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243019_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243019_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243019_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243019_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243019_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243019_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243019_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243019_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243019_at3 - -
          Human Genome U95 Set 71194_PM_at - -
          Human Genome U95 Set 71194_at - -
          Human Genome U133 Set 243019_PM_at - -
          Human Genome U133 Set 243019_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0