Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03657704_m1
          See other LOC340090 GE Assays ›
          Gene Symbol:
          LOC340090
          Gene Name:
          uncharacterized LOC340090
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 73195290 - 73201961 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03657704_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC340090
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_134291.1 1 - 2 145 90
          GenBank mRNA BC043557.1 - 1 - 2 145 90
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BI764873.1 - 1 - 2 145 Chr. 5 - 73196878 - 73201961 [-] on Build GRCh38 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transmembrane protein 174 TMEM174 -23.9kb Anti-sense Chr. 5 - 73173196 - 73175143 on NCBI Build GRCh38
          Downstream forkhead box D1 FOXD1 FKHL8,
          FREAC-4,
          FREAC4
          244.3kb Sense Chr. 5 - 73446256 - 73448527 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011882 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192435 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192389 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011969 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192457 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192452 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192470 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011893 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192376 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011956 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192473 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011890 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15710183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011957 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011949 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192476 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192460 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011917 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192417 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011887 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192466 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192478 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011911 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011856 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011916 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192487 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192372 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192380 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011931 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192416 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192418 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011922 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192483 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192388 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192475 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011865 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011891 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011947 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192392 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192401 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011951 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011952 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011926 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15710177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011862 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192409 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192370 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011871 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192393 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011908 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011858 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192479 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011910 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011945 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192410 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192390 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192378 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192453 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192436 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192455 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192450 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011909 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011924 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011958 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192447 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192429 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011929 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192407 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011968 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011859 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011869 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011961 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011889 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011902 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192434 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192484 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192412 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04011965 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192454 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192433 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24192486 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 376702 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 875720 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 105429 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 339673 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3780834 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 11785 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3780834 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01255980 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01255973 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1562492_PM_at - -
          Human Genome U133 Plus 2.0 Array 1562492_at - -
          Human Genome U133 Plus 2.0 Array 1562492_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562492_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562492_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562492_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562492_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562492_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562492_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562492_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562492_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562492_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562492_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562492_at1 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1869276 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 1780689 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_91172 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1861575 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1861575 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1869276 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0