Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03657146_s1
          See other LOC100134937 GE Assays ›
          Gene Symbol:
          LOC100134937
          Gene Name:
          uncharacterized LOC100134937
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03657146_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100134937
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC035129.2 - 1 752 75
          BC042376.1 - 1 741 75
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AW779928.1 - 1 Chr. 4 - 6985923 - 6986388 [+] on Build GRCh38 75
          GenBankmRNA AA009613.1 - 1 Chr. 4 - 6985923 - 6986395 [+] on Build GRCh38 75
          GenBankmRNA CB046708.1 - 1 639 Chr. 4 - 6986220 - 6986924 [-] on Build GRCh38 75
          GenBankmRNA AI743207.1 - 1 Chr. 4 - 6985921 - 6986453 [+] on Build GRCh38 75
          GenBankmRNA AW418879.1 - 1 Chr. 4 - 6985921 - 6986384 [+] on Build GRCh38 75
          GenBankmRNA BQ003606.1 - 1 Chr. 4 - 6985918 - 6986533 [+] on Build GRCh38 75
          GenBankmRNA AW379352.1 - 1 Chr. 4 - 6986024 - 6986377 [+] on Build GRCh38 75
          GenBankmRNA AI978565.1 - 1 Chr. 4 - 6985918 - 6986365 [+] on Build GRCh38 75
          GenBankmRNA AI041848.1 - 1 Chr. 4 - 6985921 - 6986378 [+] on Build GRCh38 75
          GenBankmRNA BU621162.1 - 1 Chr. 4 - 6985921 - 6986628 [+] on Build GRCh38 75
          GenBankmRNA BE674205.1 - 1 Chr. 4 - 6985921 - 6986417 [+] on Build GRCh38 75
          GenBankmRNA AI740939.1 - 1 Chr. 4 - 6985928 - 6986444 [+] on Build GRCh38 75
          GenBankmRNA AA812510.1 - 1 Chr. 4 - 6985921 - 6986383 [+] on Build GRCh38 75
          GenBankmRNA AI742496.1 - 1 Chr. 4 - 6985921 - 6986346 [+] on Build GRCh38 75
          GenBankmRNA DB514282.1 - 1 60 Chr. 4 - 6985925 - 6986353 [-] on Build GRCh38 75
          GenBankmRNA AI051096.1 - 1 Chr. 4 - 6985921 - 6986396 [+] on Build GRCh38 75
          GenBankmRNA BM981909.1 - 1 Chr. 4 - 6985918 - 6986592 [+] on Build GRCh38 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream TBC1 domain family member 14 TBC1D14 Anti-sense Chr. 4 - 6909444 - 7033118 on NCBI Build GRCh38
          Downstream uncharacterized LOC100129931 LOC100129931 43.5kb Sense Chr. 4 - 7030554 - 7046231 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3427239 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 7418 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3427239 - -
          Agilent Human 1B Microarray G4111A A_32_P97122 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 229685_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 240408_PM_at - -
          Human Genome U133 Plus 2.0 Array 229685_PM_at - -
          Human Genome U133 Plus 2.0 Array 240408_PM_at - -
          Human Genome U133 Plus 2.0 Array 229685_at - -
          Human Genome U133 Plus 2.0 Array 240408_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240408_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240408_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229685_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229685_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240408_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229685_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229685_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240408_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240408_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240408_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240408_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240408_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229685_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229685_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229685_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229685_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229685_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229685_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240408_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240408_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240408_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229685_at1 - -
          Human Genome U95 Set 60203_at - -
          Human Genome U95 Set 62190_r_at - -
          Human Genome U95 Set 60203_PM_at - -
          Human Genome U95 Set 62190_PM_r_at - -
          Human Genome U133 Set 240408_at - -
          Human Genome U133 Set 240408_PM_at - -
          Human Genome U133 Set 229685_at - -
          Human Genome U133 Set 229685_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7dfb8678b5-vqhv7:80/100.66.129.46:80.
          git-commit: 632cddff25488e9c1007a93c87c1899f725eef0d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0