Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000571356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000571356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000571356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000571356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000571356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000576807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000576807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000576807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
ENST00000576807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000428155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000571356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000571356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000571356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000571356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000571356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000575497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000576807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000576807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000576807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
ENST00000576807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006666.hg.1 |
NR_132322.1 |
Refseq |