Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03652958_s1
          See other ARGFXP2 GE Assays ›
          Gene Symbol:
          ARGFXP2
          Gene Name:
          arginine-fifty homeobox pseudogene 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 32150357 - 32151571 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03652958_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ARGFXP2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002222.1 1 445 76
          GenBank mRNA AA613199.1 - 1 76

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC105371730 LOC105371730 -7.2kb Sense Chr. 17 - 32141226 - 32143135 on NCBI Build GRCh38
          Downstream rhomboid like 3 RHBDL3 RHBDL4,
          VRHO
          114.6kb Anti-sense Chr. 17 - 32266167 - 32324661 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700010342.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001349.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3349289 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3738 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3737 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 28502 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3349289 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3349289 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852072 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285357 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285375 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16970383 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285378 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852083 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285365 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852082 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285355 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285374 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852085 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285362 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16970387 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852095 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285372 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285370 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852097 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852096 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285355 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285375 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285372 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285362 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852083 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285370 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852095 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285357 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285365 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852085 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285378 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285374 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852082 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16970383 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852096 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10285360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16970387 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852072 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30852097 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285315 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285312 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285326 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285321 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285329 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285325 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285359 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16970387 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285331 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285319 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16970375 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285365 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16970370 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16970384 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285328 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285377 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16970383 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285374 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285322 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285372 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285362 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285376 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285323 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285355 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285333 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285375 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10285367 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0300008530.hg.1 ENST00000334384.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300008530.hg.1 ENST00000334384.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300008530.hg.1 ENST00000334384.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300008530.hg.1 ENST00000334384.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300008530.hg.1 ENST00000334384.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300008530.hg.1 NM_001012659.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300008530.hg.1 NM_001012659.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300008530.hg.1 NM_001012659.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300008530.hg.1 NM_001012659.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300008530.hg.1 NM_001012659.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300008530.hg.1 NM_001012659.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300008530.hg.1 NM_001012659.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17017901.hg.1 NR_002222.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17017901.hg.1 TC17001349.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700010342.hg.1 ENST00000579304.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010342.hg.1 ENST00000579304.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010342.hg.1 NR_002222.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010342.hg.1 NR_002222.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010342.hg.1 NR_002222.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010342.hg.1 NR_002222.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010342.hg.1 NR_002222.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008478.hg.1 ENST00000489176.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700181211.hg.1 NR_002222.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700181211.hg.1 TC1700010342.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700181211.hg.1 NR_002222.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700181213.hg.1 NR_002222.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700181213.hg.1 TC1700010342.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700181213.hg.1 NR_002222.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700181212.hg.1 ENST00000579304.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700181212.hg.1 NR_002222.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700181212.hg.1 TC1700010342.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700181212.hg.1 ENST00000579304.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700181212.hg.1 NR_002222.1 Refseq
          Affymetrix Human Clariom D Assay TC0300008530.hg.1 ENST00000334384.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300008530.hg.1 ENST00000334384.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300008530.hg.1 ENST00000334384.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300008530.hg.1 ENST00000334384.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300008530.hg.1 ENST00000334384.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300008530.hg.1 NM_001012659.1 Refseq
          Affymetrix Human Clariom D Assay TC0300008530.hg.1 NM_001012659.1 Refseq
          Affymetrix Human Clariom D Assay TC0300008530.hg.1 NM_001012659.1 Refseq
          Affymetrix Human Clariom D Assay TC0300008530.hg.1 NM_001012659.1 Refseq
          Affymetrix Human Clariom D Assay TC0300008530.hg.1 NM_001012659.1 Refseq
          Affymetrix Human Clariom D Assay TC0300008530.hg.1 NM_001012659.1 Refseq
          Affymetrix Human Clariom D Assay TC0300008530.hg.1 NM_001012659.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700181209.hg.1 NR_002222.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700181209.hg.1 TC1700010342.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700181209.hg.1 NR_002222.1 Refseq
          Affymetrix Human Clariom D Assay TC0500008478.hg.1 ENST00000489176.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700010342.hg.1 ENST00000579304.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700010342.hg.1 ENST00000579304.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700010342.hg.1 NR_002222.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010342.hg.1 NR_002222.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010342.hg.1 NR_002222.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010342.hg.1 NR_002222.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010342.hg.1 NR_002222.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700181210.hg.1 ENST00000579304.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700181210.hg.1 NR_002222.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700181210.hg.1 TC1700010342.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700181210.hg.1 ENST00000579304.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700181210.hg.1 NR_002222.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008160 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008160 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1814353 NR_002222.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1814353 AA613199.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2095571 NR_002222.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1814353 NR_002222.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1814353 AA613199.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2095571 NR_002222.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1814353 NR_002222.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1814353 AA613199.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2095571 NR_002222.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2095571 NR_002222.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1814353 NR_002222.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1814353 AA613199.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 1450181 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_18049 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2095571 NR_002222.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2095571 NR_002222.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1814353 NR_002222.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1814353 AA613199.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2095571 NR_002222.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1814353 NR_002222.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1814353 AA613199.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1814353 NR_002222.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1814353 AA613199.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2095571 NR_002222.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7d7666c9cf-ndzlw:80/100.66.129.171:80.
          git-commit: 173eb311d069d8bb3c8868db8478582b9fe5b4f6
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0