Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03405163_g1
          See other DRD5 GE Assays ›
          Gene Symbol:
          DRD5
          Gene Name:
          dopamine receptor D5
          Gene Aliases:
          DBDR, DRD1B, DRD1L2
          Chromosome Location:
          Chr.4: 9781634 - 9784009 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03405163_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DRD5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000798.4 NP_000789.1 1 1254 87
          GenBank mRNA AK313897.1 - 1 1081 87
          AY136750.1 - 1 858 87
          BC009748.2 - 1 909 3928370 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400006853.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DRD5

          Gene Ontology Categories:

          Function(s) Process(es)

          synaptic transmission, dopaminergic
          response to amphetamine
          regulation of systemic arterial blood pressure by vasopressin
          norepinephrine-epinephrine vasoconstriction involved in regulation of systemic arterial blood pressure
          cellular calcium ion homeostasis
          adenylate cyclase-activating G protein-coupled receptor signaling pathway
          activation of adenylate cyclase activity
          adenylate cyclase-activating dopamine receptor signaling pathway
          chemical synaptic transmission
          learning
          associative learning
          transmission of nerve impulse
          negative regulation of NAD(P)H oxidase activity
          wound healing
          response to cocaine
          positive regulation of adenylate cyclase activity
          negative regulation of blood pressure
          regulation of female receptivity
          sensitization
          phospholipase C-activating dopamine receptor signaling pathway
          long-term synaptic depression
          cellular response to catecholamine stimulus
          reactive oxygen species metabolic process
          dopamine neurotransmitter receptor activity, coupled via Gs
          dopamine neurotransmitter receptor activity
          dopamine binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02919566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23071781 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23071772 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23071780 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23071783 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23071830 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02919563 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23071758 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 594970 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 62040 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 315077 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P862886 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P54274 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0062 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P862886 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0061 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P54274 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 10605 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 36832 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 26756 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 49392 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 61612 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P862886 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P54274 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P862886 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P54274 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23071758 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23071772 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23071765 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15545006 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23071780 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02919567 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02919566 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02919563 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23071781 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15545047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15544997 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23071783 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02919563 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23071780 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23071783 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23071772 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23071758 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23071765 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02919566 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23071781 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02919567 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15544997 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15545047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15545006 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P862886 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P54274 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02919565 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02919567 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02919566 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02919564 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15545035 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00917158 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P54274 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P862886 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 9177 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 2709 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 1499 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 1491 - -
          Agilent Human 1B Microarray G4111A A_32_P388188 - -
          Agilent Human 1B Microarray G4111A A_32_P54274 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15545035 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02919564 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02919568 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02919565 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_000798_at - -
          Affymetrix Human Gene 1.0 ST Array 8053726 - -
          Affymetrix Human Gene 1.0 ST Array 8053727 - -
          Affymetrix Human Gene 1.0 ST Array 7905026 - -
          Affymetrix Human Gene 1.0 ST Array 8094160 - -
          Affymetrix Human Gene 1.0 ST Array 7905024 - -
          Affymetrix Human Gene 1.0 ST Array 7905023 - -
          Affymetrix Human Gene 1.0 ST Array 8053729 - -
          Affymetrix Human Gene 1.0 ST Array 7905022 - -
          Affymetrix Human Gene 1.0 ST Array 8094159 - -
          Affymetrix Human Gene 1.0 ST Array 8053730 - -
          Affymetrix Human Gene 1.0 ST Array 8094158 - -
          Affymetrix Human Gene 1.0 ST Array 7905027 - -
          Affymetrix Human Gene 1.0 ST Array 8094157 - -
          Affymetrix Human Gene 1.1 ST Array 8094156 - -
          Affymetrix Human Gene 1.1 ST Array 7905020 - -
          Affymetrix Human Gene 1.1 ST Array 8053725 - -
          Affymetrix Human Gene 1.1 ST Array 8053728 - -
          Affymetrix Human Gene 1.1 ST Array 7905025 - -
          Affymetrix HT HG-U133+ PM Array Plate 208486_PM_at - -
          Affymetrix Human Genome U219 Array 11732695_at - -
          Affymetrix Human Genome U219 Array 11732695_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0400006853.hg.1 ENST00000304374.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006853.hg.1 ENST00000304374.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006853.hg.1 ENST00000304374.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006853.hg.1 ENST00000304374.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006853.hg.1 NM_000798.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006853.hg.1 NM_000798.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006853.hg.1 NM_000798.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006853.hg.1 NM_000798.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400006853.hg.1 NM_000798.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400147206.hg.1 ENST00000304374.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0400147206.hg.1 NM_000798.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400147206.hg.1 TC0400006853.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400147206.hg.1 NM_000798.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400147206.hg.1 ENST00000304374.3 Ensembl
          Affymetrix Human Clariom D Assay TC0400006853.hg.1 ENST00000304374.3 Ensembl
          Affymetrix Human Clariom D Assay TC0400006853.hg.1 ENST00000304374.3 Ensembl
          Affymetrix Human Clariom D Assay TC0400006853.hg.1 ENST00000304374.3 Ensembl
          Affymetrix Human Clariom D Assay TC0400006853.hg.1 ENST00000304374.3 Ensembl
          Affymetrix Human Clariom D Assay TC0400006853.hg.1 NM_000798.4 Refseq
          Affymetrix Human Clariom D Assay TC0400006853.hg.1 NM_000798.4 Refseq
          Affymetrix Human Clariom D Assay TC0400006853.hg.1 NM_000798.4 Refseq
          Affymetrix Human Clariom D Assay TC0400006853.hg.1 NM_000798.4 Refseq
          Affymetrix Human Clariom D Assay TC0400006853.hg.1 NM_000798.4 Refseq
          Human Genome U133 Plus 2.0 Array 208486_PM_at - -
          Human Genome U133 Plus 2.0 Array 208486_at - -
          Human Genome U133A 2.0 Array 208486_at - -
          Human Genome U133A 2.0 Array 208486_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_000798_at - -
          AffymetrixPathwork Diagnostics Pathchip pw596615 - -
          Human Genome U95 Set 33050_PM_at - -
          Human Genome U95 Set 33050_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208486_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208486_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208486_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208486_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208486_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208486_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208486_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208486_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208486_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208486_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208486_at5 - -
          Human Genome U133 Set 208486_at - -
          Human Genome U133 Set 208486_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A025282 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A025282 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 131671 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1689043 BC009748.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1689043 NM_000798.4 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1689043 BC009748.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1689043 NM_000798.4 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1689043 BC009748.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1689043 NM_000798.4 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1689043 BC009748.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1689043 NM_000798.4 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1689043 BC009748.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1689043 NM_000798.4 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_34328907-S BC009748.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_34328907-S NM_000798.4 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_27662-S - -
          Illumina human-6 v1.0 expression beadchip 0002120112 - -
          Illumina humanRef-8 v1.0 expression beadchip 0002120112 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1689043 BC009748.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1689043 NM_000798.4 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1689043 BC009748.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1689043 NM_000798.4 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 7560053 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_27662 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1689043 BC009748.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1689043 NM_000798.4 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1689043 BC009748.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1689043 NM_000798.4 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1689043 BC009748.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1689043 NM_000798.4 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1689043 BC009748.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1689043 NM_000798.4 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1689043 BC009748.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1689043 NM_000798.4 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5db945b66b-v5hlb:80/100.66.134.185:80.
          git-commit: 84360441031e17f3391aad9879d55c303cd12901
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0