Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03300559_s1
          See other SLED1 GE Assays ›
          Gene Symbol:
          SLED1
          Gene Name:
          proteoglycan 3 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 184798296 - 184799046 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03300559_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLED1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003542.1 1 556 88
          GenBank mRNA AY358224.1 - 1 556 88

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream centromere protein U CENPU CENP50,
          CENPU50,
          KLIP1,
          MLF1IP,
          PBIP1
          -64.2kb Sense Chr. 4 - 184694065 - 184734132 on NCBI Build GRCh38
          Downstream MIR3945 host gene MIR3945HG 44.3kb Sense Chr. 4 - 184843296 - 184855652 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400012622.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04001810.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P927716 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P927716 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 6360 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 62203 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P927716 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P927716 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845725 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684520 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845755 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684581 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845730 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684540 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684517 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684553 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684518 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684558 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845763 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684589 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684534 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684542 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845720 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684567 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845775 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845713 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845728 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684548 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684554 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845710 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845721 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684566 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845717 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684527 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684532 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684537 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845746 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684588 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684544 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845711 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845764 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845722 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684574 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845712 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845737 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684585 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845743 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684562 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845769 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684579 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845766 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684549 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684575 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684567 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845764 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845746 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684549 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684518 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684579 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845717 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845711 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845728 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684585 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845730 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845755 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684527 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845766 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845763 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684562 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684574 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684548 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845775 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845712 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684566 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684589 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845713 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684588 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684575 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684554 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845769 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684542 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845737 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684540 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684553 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845721 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684544 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845720 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684520 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684558 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684537 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684534 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684532 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684517 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03684581 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845725 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845743 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845710 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23845722 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P927716 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P927716 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684539 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684553 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684568 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684544 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684542 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684560 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684529 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684534 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684573 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684585 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684577 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684520 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684581 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684532 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684526 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684514 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684537 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684516 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684583 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684558 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684579 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684590 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684523 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684509 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684551 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684512 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684562 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684518 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684566 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684547 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684549 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03684571 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15645901 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_003542_at - -
          Affymetrix Human Gene 1.0 ST Array 8103976 - -
          Affymetrix Human Gene 1.1 ST Array 8103975 - -
          Affymetrix Human Genome U219 Array 11753498_at - -
          Affymetrix Human Genome U219 Array 11753498_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0400012622.hg.1 ENST00000506741.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012622.hg.1 ENST00000506741.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012622.hg.1 ENST00000506741.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012622.hg.1 ENST00000512051.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012622.hg.1 NR_003542.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012622.hg.1 NR_003542.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012622.hg.1 NR_003542.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04024703.hg.1 ENST00000506741.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04024703.hg.1 NR_003542.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04024703.hg.1 TC04001810.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0400186439.hg.1 ENST00000506741.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400186439.hg.1 NR_003542.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400186439.hg.1 TC0400012622.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400186439.hg.1 NR_003542.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400186439.hg.1 ENST00000506741.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400186441.hg.1 ENST00000506741.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400186441.hg.1 NR_003542.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400186441.hg.1 TC0400012622.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400186441.hg.1 ENST00000506741.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400186441.hg.1 NR_003542.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012622.hg.1 ENST00000506741.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012622.hg.1 ENST00000506741.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012622.hg.1 ENST00000506741.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012622.hg.1 ENST00000512051.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012622.hg.1 NR_003542.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012622.hg.1 NR_003542.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012622.hg.1 NR_003542.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400186440.hg.1 ENST00000506741.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400186440.hg.1 ENST00000512051.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400186440.hg.1 NR_003542.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400186440.hg.1 TC0400012622.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400186440.hg.1 ENST00000506741.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400186440.hg.1 ENST00000512051.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400186440.hg.1 NR_003542.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A025073 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A025073 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1673322 NR_003542.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1673322 AY358224.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1673322 NR_003542.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1673322 AY358224.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1673322 NR_003542.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1673322 AY358224.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1673322 NR_003542.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1673322 AY358224.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1673322 NR_003542.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1673322 AY358224.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1673322 NR_003542.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1673322 AY358224.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1673322 NR_003542.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1673322 AY358224.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0