Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03298738_s1
          See other SNORD7 GE Assays ›
          Gene Symbol:
          SNORD7
          Gene Name:
          small nucleolar RNA, C/D box 7
          Gene Aliases:
          Z30, mgU6-47
          Chromosome Location:
          Chr.17: 35573657 - 35573753 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03298738_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNORD7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003037.1 1 53 80
          GenBank mRNA AJ007733.1 - 1 53 80
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA592905.1 - 1 Chr. 17 - 35573351 - 35573758 [-] on Build GRCh38 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC105371743 LOC105371743 Sense Chr. 17 - 35568114 - 35574313 on NCBI Build GRCh38
          Downstream peroxisomal biogenesis factor 12 PEX12 PAF-3,
          PBD3A
          1.0kb Anti-sense Chr. 17 - 35574795 - 35578637 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700007598.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000398.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300605 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867509 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300622 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867520 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300604 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300609 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867512 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300607 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300612 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867500 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867515 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300616 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300600 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867511 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300610 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867510 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867507 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300602 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16972689 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16972690 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300606 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867485 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867504 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867516 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867517 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867504 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867512 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867511 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867507 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867485 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300602 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300622 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867500 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867510 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300616 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300610 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867509 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867517 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300605 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867516 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300607 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867520 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16972689 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300606 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16972690 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300600 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30867515 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300609 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300604 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10300612 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300597 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300614 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300654 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300592 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300668 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300649 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300669 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16972701 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300600 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300661 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300606 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300612 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300591 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16972693 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16972706 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16972707 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300657 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300618 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16972689 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300594 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300598 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300607 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300623 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300609 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300647 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300604 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300602 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300666 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300664 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300619 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300662 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10300616 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16972708 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_003037_at - -
          Affymetrix Human Gene 1.0 ST Array 8006541 - -
          Affymetrix Human Gene 1.1 ST Array 8006540 - -
          Affymetrix Human Transcriptome Array 2.0 PSR17004997.hg.1 ENST00000384567.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17004997.hg.1 NR_003037.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17004997.hg.1 TC17000398.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700007598.hg.1 ENST00000384567.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007598.hg.1 NR_003037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700154671.hg.1 ENST00000384567.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154671.hg.1 NR_003037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700154671.hg.1 TC1700007598.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700154671.hg.1 ENST00000384567.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154671.hg.1 NR_003037.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007598.hg.1 ENST00000384567.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007598.hg.1 NR_003037.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3236404 NR_003037.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3236404 AJ007733.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3236404 NR_003037.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3236404 AJ007733.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3236404 NR_003037.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3236404 AJ007733.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3236404 NR_003037.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3236404 AJ007733.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0