Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03297860_m1
          See other ZFAT-AS1 GE Assays ›
          Gene Symbol:
          ZFAT-AS1
          Gene Name:
          ZFAT antisense RNA 1
          Gene Aliases:
          NCRNA00070, SAS-ZFAT, ZFAT-AS, ZFATAS
          Chromosome Location:
          Chr.8: 134598071 - 134600689 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03297860_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZFAT-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002438.1 1 - 2 504 111
          GenBank mRNA AB167742.1 - 1 - 2 504 111

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC101927822 LOC101927822 -695.7kb Sense Chr. 8 - 133886496 - 133902407 on NCBI Build GRCh38
          Downstream uncharacterized LOC102723694 LOC102723694 191.3kb Anti-sense Chr. 8 - 134789112 - 134832091 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800008994.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000773.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3237005 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3772 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3773 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 50257 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3237005 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3237005 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26774655 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06492265 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06492264 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26774644 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06492262 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06492261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06492257 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06492266 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26774646 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26774657 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26774649 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26774645 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06492269 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16137927 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06492261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06492262 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06492257 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26774645 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26774657 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26774649 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06492266 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26774644 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06492265 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16137927 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06492269 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26774646 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26774655 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06492264 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06492267 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06492262 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06492268 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06492271 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06492263 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06492257 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06492273 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06492258 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06492270 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06492265 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06492260 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR08010388.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08010388.hg.1 NR_002438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08010388.hg.1 TC08000773.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08010389.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08010389.hg.1 NR_002438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08010389.hg.1 TC08000773.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0800008994.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008994.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008994.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008994.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008994.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008994.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008994.hg.1 ENST00000611506.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008994.hg.1 ENST00000613742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008994.hg.1 ENST00000620907.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008994.hg.1 NR_002438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008994.hg.1 NR_002438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008994.hg.1 NR_002438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008994.hg.1 NR_002438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008994.hg.1 NR_002438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008994.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800161359.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161359.hg.1 ENST00000613742.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161359.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800161359.hg.1 TC0800008994.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800161359.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800161359.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161359.hg.1 ENST00000613742.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008994.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008994.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008994.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008994.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008994.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008994.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008994.hg.1 ENST00000611506.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008994.hg.1 ENST00000613742.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008994.hg.1 ENST00000620907.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008994.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008994.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008994.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008994.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008994.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008994.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800161360.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161360.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800161360.hg.1 TC0800008994.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800161360.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161360.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800161363.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161363.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800161363.hg.1 TC0800008994.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800161363.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161363.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800161358.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161358.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800161358.hg.1 TC0800008994.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800161358.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161358.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800161362.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161362.hg.1 ENST00000611506.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161362.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800161362.hg.1 TC0800008994.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800161362.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161362.hg.1 ENST00000611506.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161362.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800161357.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161357.hg.1 ENST00000620907.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161357.hg.1 NR_002438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800161357.hg.1 TC0800008994.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800161357.hg.1 ENST00000505776.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161357.hg.1 ENST00000620907.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800161357.hg.1 NR_002438.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036948 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036948 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1723761 NR_002438.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1723761 AB167742.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2177575 NR_002438.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2177575 AB167742.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1723761 NR_002438.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1723761 AB167742.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2177575 NR_002438.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2177575 AB167742.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1723761 NR_002438.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1723761 AB167742.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2177575 NR_002438.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2177575 AB167742.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2177575 NR_002438.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2177575 AB167742.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1723761 NR_002438.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1723761 AB167742.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 6550477 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_28847 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2177575 NR_002438.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2177575 AB167742.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2177575 NR_002438.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2177575 AB167742.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1723761 NR_002438.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1723761 AB167742.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2177575 NR_002438.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2177575 AB167742.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1723761 NR_002438.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1723761 AB167742.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1723761 NR_002438.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1723761 AB167742.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2177575 NR_002438.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2177575 AB167742.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0