Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03297272_sH
          See other ANXA2P1 GE Assays ›
          Gene Symbol:
          ANXA2P1
          Gene Name:
          annexin A2 pseudogene 1
          Gene Aliases:
          ANX2L1, ANX2P1, LPC2A
          Chromosome Location:
          Chr.4: 153307469 - 153308811 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03297272_sH
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ANXA2P1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_001562.2 1 1151 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream FH2 domain containing 1 FHDC1 -327.8kb Anti-sense Chr. 4 - 152911570 - 152979696 on NCBI Build GRCh38
          Downstream meiotic nuclear divisions 1 MND1 GAJ 35.8kb Anti-sense Chr. 4 - 153344649 - 153415097 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P204244 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P204244 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 22752 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 6863 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P204244 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P204244 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15628275 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699130 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537422 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699105 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699131 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15628279 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699101 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699104 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537409 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537451 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699122 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699141 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699113 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699144 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537424 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699107 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699140 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699116 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537444 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537458 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537416 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699110 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699129 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699108 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699132 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699127 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699102 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537406 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699145 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537407 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699145 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537458 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699130 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699116 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699102 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699129 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699105 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537409 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537444 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15628279 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699107 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537451 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699101 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537416 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699141 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699122 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15628275 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699113 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699104 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699108 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699132 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537407 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699131 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699140 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537424 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699110 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699127 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537422 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23699144 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03537406 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P204244 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P204244 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537422 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537437 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537417 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537445 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537415 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537416 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537411 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537409 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537407 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537443 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537425 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537419 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537462 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537454 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15628281 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537441 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537458 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537430 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537456 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537439 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537435 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03537451 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_001562_at - -
          Affymetrix Human Gene 1.0 ST Array 8103241 - -
          Affymetrix Human Gene 1.1 ST Array 8103240 - -
          Affymetrix HT HG-U133+ PM Array Plate 210876_PM_at - -
          Affymetrix Human Genome U219 Array 11751392_at - -
          Affymetrix Human Genome U219 Array 11751392_PM_at - -
          Human Genome U133 Plus 2.0 Array 210876_at - -
          Human Genome U133 Plus 2.0 Array 210876_PM_at - -
          Human Genome U133A 2.0 Array 210876_at - -
          Human Genome U133A 2.0 Array 210876_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NR_001562_at - -
          AffymetrixPathwork Diagnostics Pathchip pw567509 - -
          Human Genome U95 Set 31684_at - -
          Human Genome U95 Set 31444_s_at - -
          Human Genome U95 Set 31684_PM_at - -
          Human Genome U95 Set 769_s_at - -
          Human Genome U95 Set 31444_PM_s_at - -
          Human Genome U95 Set 769_PM_s_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210876_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210876_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210876_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210876_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210876_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210876_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210876_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210876_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210876_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210876_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210876_at5 - -
          Human Genome U133 Set 210876_PM_at - -
          Human Genome U133 Set 210876_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024871 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024870 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024870 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024871 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1698100 NR_001562.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2041101 NR_001562.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1698100 NR_001562.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2041101 NR_001562.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1698100 NR_001562.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2041101 NR_001562.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2041101 NR_001562.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1698100 NR_001562.2 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 4200474 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_10494 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2041101 NR_001562.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1698100 NR_001562.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2041101 NR_001562.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2041101 NR_001562.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1698100 NR_001562.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1698100 NR_001562.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2041101 NR_001562.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-7p7bh:80/100.66.135.138:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0