Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000421269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000421269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000421269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000517575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000517575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000517575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000518395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000518395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000524171.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000524171.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000524171.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000524171.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000524171.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000593851.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000593851.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000593851.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
ENST00000593851.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001102609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001102609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001102609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001102609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305393.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305393.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305393.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305393.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305394.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305394.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305394.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305394.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NM_001305397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NR_131091.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NR_131091.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NR_131091.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NR_131091.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NR_131091.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NR_131092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NR_131092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NR_131092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009392.hg.1 |
NR_131092.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05013570.hg.1 |
ENST00000517575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05013570.hg.1 |
ENST00000521850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05013570.hg.1 |
NM_001102609.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05013570.hg.1 |
TC05000933.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
ENST00000421269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
ENST00000517575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
ENST00000524171.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
ENST00000593851.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NM_001102609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NM_001305393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NM_001305394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NM_001305395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NM_001305396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NM_001305397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NR_131091.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NR_131092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
TC0500009392.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
ENST00000524171.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
ENST00000421269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
ENST00000517575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
ENST00000593851.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NM_001102609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NM_001305393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NM_001305394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NM_001305395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NM_001305396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NM_001305397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NR_131091.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165797.hg.1 |
NR_131092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000421269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000421269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000421269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000517575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000517575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000517575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000518395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000518395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000524171.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000524171.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000524171.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000524171.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000524171.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000593851.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000593851.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000593851.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
ENST00000593851.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001102609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001102609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001102609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001102609.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NM_001305397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NR_131091.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NR_131091.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NR_131091.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NR_131091.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NR_131091.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NR_131092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NR_131092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NR_131092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009392.hg.1 |
NR_131092.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165792.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500165792.hg.1 |
NM_001305393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165792.hg.1 |
NM_001305394.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165792.hg.1 |
TC0500009392.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500165792.hg.1 |
ENST00000521850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500165792.hg.1 |
NM_001305393.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500165792.hg.1 |
NM_001305394.1 |
Refseq |